Skip to Content
Merck
All Photos(1)

Key Documents

51475

Sigma-Aldrich

Yeast

lyticase substrate, powder

Synonym(s):

Dry brewer′s yeast

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

product name

Yeast, powder, slightly brown

form

powder

Quality Level

color

slightly brown

application(s)

food and beverages

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Yeast is an inactive edible single-celled organism widely used in the food industry for baking, winemaking, and brewing. This product is dried irrespective of enzyme activity. Yeast serves as a substrate for lyticase.

Application

Yeast has been used:
  • as a structure modifier to alter the morphology and rheology of phase-separated biopolymer gels
  • to induce hyperthermia in rats to perform antipyretic activity test
  • in the extraction and enrichment method for the metaproteomic characterization of dissolved organic matter in groundwater samples

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hilkatul Ilmi et al.
Evidence-based complementary and alternative medicine : eCAM, 2021, 8848797-8848797 (2021-03-23)
To determine the analgesic and antipyretic activities of a tablet derived from Andrographis paniculata ethyl acetate fraction (AS201-01) in animal models. The tablet derived from AS201-01 contains an equivalent of 35 mg andrographolide per tablet. Analgesic activity was determined using an
Hilkatul Ilmi et al.
Evidence-based complementary and alternative medicine : eCAM, 2021, 8848797-8848797 (2021-03-23)
To determine the analgesic and antipyretic activities of a tablet derived from Andrographis paniculata ethyl acetate fraction (AS201-01) in animal models. The tablet derived from AS201-01 contains an equivalent of 35 mg andrographolide per tablet. Analgesic activity was determined using an
Anna Laura Capriotti et al.
Journal of separation science, 44(8), 1612-1620 (2020-11-26)
Metaproteomic analysis of aquifer systems provides valuable information on the microbial populations, their influence on drinking water quality, and the effect on human health. In the present paper, an extraction and enrichment method by C18 extra-wide pore cartridge was developed
Anna Laura Capriotti et al.
Journal of separation science, 44(8), 1612-1620 (2020-11-26)
Metaproteomic analysis of aquifer systems provides valuable information on the microbial populations, their influence on drinking water quality, and the effect on human health. In the present paper, an extraction and enrichment method by C18 extra-wide pore cartridge was developed
Gerrit A Stuivenberg et al.
mSphere, 7(6), e0044622-e0044622 (2022-11-03)
Renal impairment associated with chronic kidney disease (CKD) causes the buildup of uremic toxins that are deleterious to patient health. Current therapies that manage toxin accumulation in CKD offer an incomplete therapeutic effect against toxins such as para-cresol (p-cresol) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service