Skip to Content
Merck
All Photos(2)

Key Documents

W102

Sigma-Aldrich

(R)-(+)-WIN 55,212-2 mesylate salt

≥98% (HPLC), powder, CB2 agonist

Synonym(s):

(R)-(+)-[2,3-Dihydro-5-methyl-3[(4-morpholinyl)methyl]pyrrolo[1,2,3-de]-1,4-benzoxazinyl]-(1-naphthalenyl)methanone mesylate salt, WIN 55212-2 methanesulfonate, WIN 552122 mesylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H26N2O3 · CH3SO3H
CAS Number:
Molecular Weight:
522.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

(R)-(+)-WIN 55,212-2 mesylate salt, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

0.1 M HCl: 0.25 mg/mL
DMSO: 12 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2.4 mg/mL
0.1 M NaOH: insoluble
H2O: insoluble

SMILES string

CS(O)(=O)=O.Cc1c(C(=O)c2cccc3ccccc23)c4cccc5OC[C@@H](CN6CCOCC6)n1c45

InChI

1S/C27H26N2O3.CH4O3S/c1-18-25(27(30)22-9-4-7-19-6-2-3-8-21(19)22)23-10-5-11-24-26(23)29(18)20(17-32-24)16-28-12-14-31-15-13-28;1-5(2,3)4/h2-11,20H,12-17H2,1H3;1H3,(H,2,3,4)/t20-;/m1./s1

InChI key

FSGCSTPOPBJYSX-VEIFNGETSA-N

Gene Information

General description

WIN 55,212-2 is a member of the aminoalkylindole class of compounds.

Application

(R)-(+)-WIN 55,212-2 mesylate salt has been used as a high affinity cannabinoid agonist for intraperitoneal injection into rats to induce behavioural sensitization. It has also been used as a synthetic cannabinoid drug and cannabinoid receptor 1 (CNR1) agonist to treat Becn2+/− mice and wild-type (WT) littermate to analyze the levels of CNR1 in the brain.

Biochem/physiol Actions

It is known to decrease the lipopolysaccharide mediated release of tumor necrosis factor- α and interleukin-1 in mice.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

The solubility of this chemical in DMF has not been determined. However, various sources state this chemical can be solubilized in about 30 mg/ml of DMF.

Legal Information

Sold with the permission of Sterling-Winthrop, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rational design and synthesis of an orally active indolopyridone as a novel conformationally constrained cannabinoid ligand possessing antiinflammatory properties
Wrobleski ST, et al.
Journal of Medicinal Chemistry, 46(11), 2110-2116 (2003)
Dual Role of PPAR-c in Induction and Expression of Behavioral Sensitization to Cannabinoid Receptor Agonist WIN55,212-2
Enayatfard L, et al.
Neuromolecular Medicine, 15, 523-535 (2013)
Autophagy activation by novel inducers prevents BECN2-mediated drug tolerance to cannabinoids
Kuramoto K, et al.
Autophagy, 12(9), 1460-1471 (2016)
José F Oliveira da Cruz et al.
Addiction biology, 26(3), e12940-e12940 (2020-08-04)
Synthetic cannabinoids have emerged as novel psychoactive substances with damaging consequences for public health. They exhibit high affinity at the cannabinoid type-1 (CB1 ) receptor and produce similar and often more potent effects as other CB1 receptor agonists. However, we
Gaëtan Philippot et al.
Toxicological sciences : an official journal of the Society of Toxicology, 166(1), 203-212 (2018-08-31)
Acetaminophen (AAP; also known as paracetamol) is the most used and only recommended analgesic and antipyretic among pregnant women and young children. However, recent findings in both humans and rodents suggest a link between developmental exposure to AAP and adverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service