Skip to Content
Merck
All Photos(1)

Documents

A6478

Sigma-Aldrich

(R,S)-AM1241

≥98% (HPLC), solid

Synonym(s):

(R,S)-3-(2-Iodo-5-nitrobenzoyl)-1-(1-methyl-2-piperidinylmethyl)-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22IN3O3
CAS Number:
Molecular Weight:
503.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

yellow

solubility

DMSO: ~18 mg/mL at 60 °C

SMILES string

CN1CCCCC1Cn2cc(C(=O)c3cc(ccc3I)[N+]([O-])=O)c4ccccc24

InChI

1S/C22H22IN3O3/c1-24-11-5-4-6-16(24)13-25-14-19(17-7-2-3-8-21(17)25)22(27)18-12-15(26(28)29)9-10-20(18)23/h2-3,7-10,12,14,16H,4-6,11,13H2,1H3

InChI key

ZUHIXXCLLBMBDW-UHFFFAOYSA-N

Application

(R,S)-AM1241 has been used as a cannabinoid CB2 agonist:
  • to study its inhibitory effect on bone cancer-induced pain and bone loss
  • to study the effect of its interaction with 17βestradiol on proliferation activity in primary human osteoblasts
  • to evaluate the sites of CB2 mediated antinociception in vivo.

Biochem/physiol Actions

AM1241 acts as an antinociceptive agent in several animal pain models. It has a potential to delay disease progression in amyotrophic lateral sclerosis (ALS) mouse model. Intrathecal, intravenous or intraperitoneal administration of AM1241 reduces hyperalgesia and allodynia in neuropathic rats.
Selective CB2 cannabinoid receptor agonist

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AM1241, a cannabinoid CB2 receptor selective compound, delays disease progression in a mouse model of amyotrophic lateral sclerosis
Kim K, et al.
European Journal of Pharmacology, 542(1-3), 100-105 (2006)
A synergistic interaction of 17-$\beta$-estradiol with specific cannabinoid receptor type 2 antagonist/inverse agonist on proliferation activity in primary human osteoblasts
Hojnik M, et al.
Biomedical Reports, 3(4), 554-558 (2015)
The cannabinoid CB2 receptor-specific agonist AM1241 increases pentylenetetrazole-induced seizure severity in Wistar rats
de Carvalho CR, et al.
Epilepsy Research, 127(2), 160-167 (2016)
Elizabeth J Rahn et al.
Pharmacology, biochemistry, and behavior, 98(4), 493-502 (2011-03-09)
Cannabinoid CB(2) agonists produce antinociception without central nervous system (CNS) side-effects. This study was designed to characterize the pharmacological and antinociceptive profile of AM1710, a CB(2) agonist from the cannabilactone class of cannabinoids. AM1710 did not exhibit off-target activity at
Immunofluorescent spectral analysis reveals the intrathecal cannabinoid agonist, AM1241, produces spinal anti-inflammatory cytokine responses in neuropathic rats exhibiting relief from allodynia
Wilkerson JL, et al.
Brain and Behavior, 2(2), 155-177 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service