Skip to Content
Merck
All Photos(5)

Key Documents

238813

Sigma-Aldrich

(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

97%

Synonym(s):

Trolox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O4
CAS Number:
Molecular Weight:
250.29
Beilstein:
1384051
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

storage condition

under inert gas

mp

187-189 °C (lit.)

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

Cc1c(C)c2OC(C)(CCc2c(C)c1O)C(O)=O

InChI

1S/C14H18O4/c1-7-8(2)12-10(9(3)11(7)15)5-6-14(4,18-12)13(16)17/h15H,5-6H2,1-4H3,(H,16,17)

InChI key

GLEVLJDDWXEYCO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid (Trolox) exhibits radical scavenging and antioxidant activities.[1][2][3][4]

Application

(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid (Trolox) has been used in 6-hydroxy-2,5,7,8-tetramethyl chromane-2-carboxylic acid equivalent antioxidant assay (TEAC) to determine the total antioxidant capacity of different fruit species[5].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Giulivi et al.
Archives of biochemistry and biophysics, 303(1), 152-158 (1993-05-15)
The reactivity of Trolox C, a water-soluble analog of vitamin E, toward protein radicals formed during the oxidation of metmyoglobin and the therefrom derived molecular products was examined in terms of the ability of the phenolic antioxidant to prevent specific
Gláucia E M L Siervo et al.
Biology of reproduction, 100(1), 112-122 (2018-07-17)
Spermatogenesis and steroidogenesis are not fully established during puberty. Especially during this period, children and adolescents may be chronically sleep deprived due to early school hours and constant exposure to artificial light and interactive activities. We have previously shown that
R Lakshmi Sundaram et al.
Journal of ethnopharmacology, 241, 111962-111962 (2019-05-24)
Spermacoce hispida (S.hispida), a potential medicinal plant has been traditionally used as an antibacterial, antieczemic, antihypertensive, antidiabetic and antihyperlipidemic agent. Although, this plant has been claimed to protect against oxidative injury and inflammatory conditions in recent studies, its cardioprotective effect
Jessica Scalzo et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(2), 207-213 (2005-02-23)
We wanted to determine the total antioxidant capacity (6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid equivalent antioxidant assay; TEAC) of different species and cultivars (strawberry and apple) and to characterize the TEAC among fruits of the same varieties (peach and apricot) but grafted on different
Dalia Mazor et al.
Biochemical and biophysical research communications, 349(3), 1171-1175 (2006-09-15)
The antioxidant properties of Bucillamine (BUC), a di-thiol compound used for treatment of rheumatoid arthritis (RA) and its possible mode of action, were investigated. BUC exhibits potent antioxidant activity similar to those of trolox and ascorbic acid. It reduces the

Questions

1–9 of 9 Questions  
  1. for this product, (±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid (Trolox), what is the stability if keep in 4-8 degree. can we keep in-20

    1 answer
    1. The recommended storage temperature of this product is 2-8°C. The item is not assigned an expiration date, but it periodically retested to assure quality. The assigned retest date is lot specific and may be found in the product Certificate of Analysis. Note that this material is packaged under inert gas. Exposure to air or moisture may accelerate degradation. The stability of this product at freezer temperatures has not been evaluated.

      Helpful?

  2. Bonjour, est-ce que le trolox est soluble dans l'eau svp?

    1 answer
    1. This product has not been tested for solubility in water. Various sources suggest that this material is poorly soluble in water at 0.5 mg/mL. The solubility reportedly increases to 3 mg/mL in PBS at pH 7.2.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  5. What is the stability of Trolox in solution? And at what temperature?

    1 answer
    1. Unfortunately, the solution stability is not determined for this product.

      Helpful?

  6. What is the molar absorption coefficient of Trolox ?

    1 answer
    1. The molar absorption coefficient is not determined for this product.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  9. What is the solubility of Product 238813, (±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid?

    1 answer
    1. Product 238813, (±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, has been reported to be soluble in DMSO, ethanol and dimethyl formamide. The solubility in these solvents is ~20mg/ml.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service