Skip to Content
Merck
All Photos(2)

Key Documents

T0678

Sigma-Aldrich

Anti-Tryptophan Hydroxylase Antibody

mouse monoclonal, WH-3

Synonym(s):

Anti-TPH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-Tryptophan Hydroxylase antibody produced in mouse, clone WH-3, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

WH-3, monoclonal

mol wt

antigen 55 kDa

contains

15 mM sodium azide

species reactivity

human, monkey, rabbit, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1:1,000 using rabbit pineal gland extract

isotype

IgG3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-Tryptophan Hydroxylase (mouse IgG3 isotype) is derived from the WH-3 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Tryptophan hydroxylase (TPH; L-tryptophan tetrahydropteridine:oxygen oxidoreductase; EC 1.14.16.4), the rate-limiting enzyme for the synthesis of serotonin, is a very specific enzyme found only inside serotoninergic neurons.
The tryptophan hydroxylase 1 gene (TPH1) is a 444 amino acid protein encoded by the gene spanning 29 kbp with11 exons on human chromosome 11p15.3-p14. The enzyme is expressed in the gut, spleen, thymus, pituitary and also in the pineal gland.

Specificity

This antibody reacts specifically with tryptophan hydroxylase.

Immunogen

recombinant rabbit tryptophan hydroxylase.

Application

Monoclonal Anti-Tryptophan Hydroxylase antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunohistochemistry
  • immunoblot
  • immunocytochemistry

Biochem/physiol Actions

The tryptophan hydroxylase 1 gene (TPH1) catalyzes the synthesis of 5-hydroxytryptophan, which is a precursor to the neurotransmitter serotonin. Variation in the gene expression affects prefrontal cortex activation during response inhibition. Mutation in the TPH1 gene leads to schizophrenia, irritable bowel syndrome (IBS) in women, suicidal behavior and depressive disorders. The encoded protein plays a vital role in regulation of cardiovascular function.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kadri Andre et al.
BMC psychiatry, 13, 118-118 (2013-04-20)
In major depression, one of the candidate genes possibly affecting the risk and severity of symptoms has been found to be tryptophan hydroxylase (TPH1). Variation in treatment response to antidepressive agents according to TPH1 genotype has also been found in
Honghai Zhang et al.
Epilepsia, 57(8), 1228-1235 (2016-06-16)
The DBA/1 mouse is a relevant animal model of sudden unexpected death in epilepsy (SUDEP), as it exhibits seizure-induced respiratory arrest (S-IRA) evoked by acoustic stimulation, followed by cardiac arrhythmia and death. Defects in serotonergic neurotransmission may contribute to S-IRA.
Brain tryptophan hydroxylase: purification of, production of antibodies to, and cellular and ultrastructural localization in serotonergic neurons of rat midbrain
Joh TH, et al.
Proceedings of the National Academy of Sciences of the USA, 72(9), 3575-3579 (1975)
Anthony C Ruocco et al.
Neuropsychology, 30(1), 18-27 (2015-12-29)
The tryptophan hydroxylase 1 gene (TPH1) catalyzes the formation of 5-hydroxytryptophan, a precursor to the neurotransmitter serotonin. Variations in the gene encoding this enzyme may underlie difficulties in impulse control; however, the proximate relationship between risk alleles for polymorphisms in
Neuronal loss in the rostral ventromedial medulla in a rat model of neuropathic pain
Leong ML, et al.
The Journal of Neuroscience, 31(47), 17028-17039 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service