Skip to Content
Merck
All Photos(1)

Documents

AB1541

Sigma-Aldrich

Anti-Tryptophan Hydroxylase Antibody

Chemicon®, from sheep

Synonym(s):

Tryptophan 5-monooxygenase 1, indoleacetic acid-5-hydroxylase, tryptophan 5-hydroxylase, tryptophan 5-monooxygenase, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TPH1(7166)

General description

Tryptophan hydroxylase (TPH) is an enzyme involved in the synthesis of the neurotransmitter serotonin. It is responsible for addition of the -OH group (hydroxylation) to the 5 position to form the amino acid 5-hydroxytryptophan (5-HTP), which is the initial and rate-limiting step in the synthesis of the hormone serotonin. It is also the first enzyme in the synthesis of melatonin.

Specificity

Reacts with human and rat brain tissues by immunoblotting and immunohistochemistry. Based on the relatively high degree of homology of tryptophan hydroxylase, the antibody should cross-react with other mammalian species.
Recognizes Tryptophan Hydroxylase. Reacts with human and rat brain tissues by immunoblotting and immunohistochemistry. Based on the relatively high degree of homology of tryptophan hydroxylase, the antibody should cross-react with other mammalian species. Recently studies have indicated that AB1541 can cross react with Tyrosine hydroxylase on western blots, weakly. Cross reactivity in the native state is untested.

Immunogen

Recombinant rabbit tryptophan hydroxylase, isolated as inclusion bodies from E. coli and purified by preparative SDS-PAGE.

Application

Detect Tryptophan Hydroxylase using this Anti-Tryptophan Hydroxylase Antibody validated for use in IH & WB.
Immunohistochemistry(peroxidase):
Performed on formaldehyde fixed tissue of a previous lot. Useful for cell body but not fiber staining.

Optimal working dilutions must be determined by the end user.

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected Tryptophan Hydroxylase on 10 μg of PC12 lysates.

Target description

55 kDa

Linkage

Replaces: AB1556

Physical form

Purified sheep polyclonal liquid in 150 mM NaCl, 10 mM HEPES (pH 7.5), 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Stable for 6 months at -20ºC to -80ºC in undiluted aliquots from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
Brain tissue

PC12 lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla Frare et al.
Frontiers in physiology, 9, 1747-1747 (2019-01-09)
Thermoregulation is necessary to maintain energy homeostasis. The novel discovery of brown adipose tissue (BAT) in humans has increased research interests in better understanding BAT thermogenesis to restore energy balance in metabolic disorders. The hibernating Arctic ground squirrel (AGS) offers
S J Bang et al.
Neuroscience, 179, 1-8 (2011-02-01)
Adolescence is a critical vulnerable period during which exposure to nicotine greatly enhances the possibility to develop drug addiction. Growing evidence suggests that serotonergic (5-HT) neurotransmission may contribute to the initiation and maintenance of addictive behavior. As the dorsal raphe
Olivier Berton et al.
Neuron, 55(2), 289-300 (2007-07-21)
We analyzed the influence of the transcription factor DeltaFosB on learned helplessness, an animal model of affective disorder wherein a subset of mice exposed to inescapable stress (IS) develop a deficit in escape behavior. Repeated IS induces DeltaFosB in the
Extrasynaptic glycine receptors of rodent dorsal raphe serotonergic neurons: a sensitive target for ethanol.
Maguire, EP; Mitchell, EA; Greig, SJ; Corteen, N; Balfour, DJ; Swinny, JD; Lambert, JJ; Belelli, D
Neuropsychopharmacology null
Chris M Panzini et al.
Journal of neurochemistry, 143(6), 708-721 (2017-09-28)
In humans a chromosomal hemideletion of the 16p11.2 region results in variable neurodevelopmental deficits including developmental delay, intellectual disability, and features of autism spectrum disorder (ASD). Serotonin is implicated in ASD but its role remains enigmatic. In this study we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service