Skip to Content
Merck
All Photos(1)

Key Documents

T1299

Sigma-Aldrich

Monoclonal Anti-Tyrosine Hydroxylase antibody produced in mouse

clone TH-2, ascites fluid

Synonym(s):

Anti-Tyrosine Hydroxylase, Mouse Anti-Tyrosine Hydroxylase, Tyrosine Hydroxylase Detection

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

TH-2, monoclonal

contains

15 mM sodium azide

species reactivity

human, monkey, guinea pig, rat, sheep, bovine, rabbit

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable using native or SDS-denatured TH.
western blot: 1:10,000 using extracts of cultured rat adrenal pheochromocytoma (PC12) cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TH(7054)
rat ... Th(25085)

General description

Monoclonal Anti-Tyrosine Hydroxylase (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Tyrosine hydroxylase (TH) RNA undergoes alternative splicing resulting in four isoforms of TH subunit protein in humans. The four human isoforms are TH-1 (55.6 kDa), TH-2 (56 kDa), TH-3 (58.1 kDa), and TH-4 (58.5 kDa). Only humans express all four isoforms.
Tyrosine hydroxylase (TH) is encoded by the gene mapped to human chromosome 11p15. TH is a tetramer of four identical subunits, which is characterized with a regulatory, catalytic, and tetramerization domains. The enzyme utilizes tyrosine, tetrahydrobiopterin (BH4) and O2 as cosubstrates, and Fe2+ as a cofactor.

Specificity

The antibody recognizes an epitope present in the N-terminal region (approx. aa 9-16) of both rodent and human tyrosine hydroxylase. Clone TH-2 reacts with the intact TH subunits.

Immunogen

rat tyrosine hydroxylase (TH).

Application

Monoclonal Anti-Tyrosine Hydroxylase antibody produced in mouse has also been used in
  • western blotting
  • indirect immunofluorescence
  • immunostaining
  • immunohistochemistry
  • enzyme linked immunosorbent assay (ELISA)
  • dot blot
  • immunoprecipitation

Monoclonal Anti-tyrosine hydroxylase antibody produced in mouse has been used in western blotting and immunocytochemistry.

Biochem/physiol Actions

Tyrosine hydroxylase (TH) catalyzes the first rate limiting step in the biosynthesis of catecholamine neurotransmitter i.e. the conversion of L-tyrosine to L-dopa. Inhibition of TH by L-phenylalanine, might play a crucial role in phenylketonuria and block the synthesis of norepinephrine. Activity of TH can be regulated by phosphorylation. Decreased expression of TH is associated with various neuropsychiatric diseases such as schizophrenia and Parkinson′s disease (PD).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical sympathectomy-induced changes in TH-, VIP-, and CGRP-immunoreactive fibers in the rat mandible periosteum: Influence on bone resorption
Cherruau M, et al.
Journal of Cellular Physiology, 194(3), 341-348 (2003)
Essential role of mesolimbic brain-derived neurotrophic factor in chronic social stress-induced depressive behaviors
Koo JW, et al.
Biological Psychiatry, 80(6), 469-478 (2016)
Induction of neurite outgrowth in PC12 cells by the bacterial nucleoside N6-methyldeoxyadenosine is mediated through adenosine A2a receptors and via cAMP and MAPK signaling pathways
Charles MP, et al.
Biochemical and biophysical research communications, 304(4), 795-800 (2003)
Multiple genes on chromosome 7 regulate dopaminergic amacrine cell number in the mouse retina
Whitney IE, et al.
Investigative Ophthalmology & Visual Science, 50(5), 1996-2003 (2009)
Human tyrosine hydroxylase isoforms inhibition by excess tetrahydropterin and unusual behavior of isoform 3 after cAMP-dependent protein kinase phosphorylation
Alterio J, et al.
Test, 273(17), 10196-10201 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service