Skip to Content
Merck
All Photos(1)

Documents

SRP3061

Sigma-Aldrich

IFN-omega human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IFN alpha II-1, IFNW1, Interferon-alpha II-1, Interferon-omega

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.01 ng/mL ED50

mol wt

19.9 kDa

packaging

pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IFNW1(3467)

General description

IFN-ω is a type I interferon, which can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-α, IFN-β, and IFN-ω, signal through IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities.  IFN-ω exhibits about 75% sequence homology with IFN- α, and contains two conserved disulfide bonds, which are necessary for full biological activity. Recombinant Human IFN-ω is a 19.9 kDa protein consisting of 173 amino acid residues.

Biochem/physiol Actions

IFN-ω is a type I interferon, which can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-α, IFN-β, and IFN-ω, signal through IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities.  IFN-ω exhibits about 75% sequence homology with IFN- α, and contains two conserved disulfide bonds, which are necessary for full biological activity. Recombinant Human IFN-ω is a 19.9 kDa protein consisting of 173 amino acid residues.

Sequence

MCDLPQNHGL LSRNTLVLLH QMRRISPFLC LKDRRDFRFP QEMVKGSQLQ KAHVMSVLHE MLQQIFSLFH TERSSAAWNM TLLDQLHTGL HQQLQHLETC LLQVVGEGES AGAISSPALT LRRYFQGIRV YLKEKKYSDC AWEVVRMEIM KSLFLSTNMQ ERLRSKDRDL GSS

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Bastard et al.
Science immunology, 6(62) (2021-08-21)
Circulating autoantibodies (auto-Abs) neutralizing high concentrations (10 ng/mL, in plasma diluted 1 to 10) of IFN-α and/or -ω are found in about 10% of patients with critical COVID-19 pneumonia, but not in subjects with asymptomatic infections. We detect auto-Abs neutralizing
Angélique Chauvineau-Grenier et al.
Journal of clinical immunology, 42(3), 459-470 (2022-01-28)
Recent studies reported the presence of pre-existing autoantibodies (auto-Abs) neutralizing type I interferons (IFNs) in at least 15% of patients with critical COVID-19 pneumonia. In one study, these auto-Abs were found in almost 20% of deceased patients across all ages.
David Goncalves et al.
Clinical & translational immunology, 10(8), e1327-e1327 (2021-08-26)
Impairment of type I interferon (IFN-I) immunity has been reported in critically ill COVID-19 patients. This defect can be explained in a subset of patients by the presence of circulating autoantibodies (auto-Abs) against IFN-I. We set out to improve the
Romain Arrestier et al.
Annals of intensive care, 12(1), 121-121 (2023-01-01)
Auto-antibodies (auto-Abs) neutralizing type I interferons (IFN) have been found in about 15% of critical cases COVID-19 pneumonia and less than 1% of mild or asymptomatic cases. Determining whether auto-Abs influence presentation and outcome of critically ill COVID-19 patients could
Quentin Philippot et al.
Journal of clinical immunology, 43(6), 1093-1103 (2023-05-20)
Autoantibodies (auto-Abs) neutralizing type I interferons (IFNs) are found in the blood of at least 15% of unvaccinated patients with life-threatening COVID-19 pneumonia. We report here the presence of auto-Abs neutralizing type I IFNs in the bronchoalveolar lavage (BAL) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service