Skip to Content
Merck

SML2284

Mito A iodide

≥98% (HPLC)

Synonym(s):

3-(4’-Azidobenzamido)-prop-1-yl]triphenylphosphonium iodide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H26IN4OP
Molecular Weight:
592.41
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=C(N=[N+]=[N-])C=C4.[I-]

InChI

1S/C28H25N4OP.HI/c29-32-31-24-19-17-23(18-20-24)28(33)30-21-10-22-34(25-11-4-1-5-12-25,26-13-6-2-7-14-26)27-15-8-3-9-16-27;/h1-9,11-20H,10,21-22H2;1H

InChI key

DMRSJCIHHULJSH-UHFFFAOYSA-N

Biochem/physiol Actions

MitoA is selective hydrogen sulfide (H2S) probe targeting mithchondria suitable for in vivo use. MitoA accumulates in mitochondria within tissues, where it is converted to MitoN by reaction with H2S. Both compounds are detected sensitively by LC-MS/MS.
selective hydrogen sulfide (H2S) probe targeting mithchondria suitable for in vivo use.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library