Skip to Content
Merck
All Photos(1)

Key Documents

SML3730

Sigma-Aldrich

Testosterone enanthate

≥98% (HPLC)

Synonym(s):

(17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 4-Androstene-17β-ol-3-one enanthate, Heptanoic acid ester with testosterone, Testosterone heptanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule III

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

SMILES string

O([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@]4(CCC(=O)C=C4CC3)C)CC2)CC1)C)C(=O)CCCCCC

InChI key

VOCBWIIFXDYGNZ-IXKNJLPQSA-N

Biochem/physiol Actions

Prodrug of testosterone that has a long elimination half-life; androgen and anabolic–androgenic steroid that acts as agonist of the androgen receptor
Testosterone enanthate is a prodrug of testosterone that has a long elimination half-life. It is an androgen and anabolic–androgenic steroid that acts as agonist of the androgen receptor (AR). Testosterone enanthate is mainly used in the treatment of low testosterone levels in men. Also, it was approved for treatment of hypogonadism in men, delayed puberty in boys, and breast cancer in women.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Testosterone undecanoate and testosterone enanthate injections are both effective and safe in transmen over 5 years of administration
Clinical Endocrinology, 89(6), 878-886 (2018)
Locomotor training with adjuvant testosterone preserves cancellous bone and promotes muscle plasticity in male rats after severe spinal cord injury
Journal of Neuroscience Research, 98(5), 843-868 (2020)
Pharmacokinetics, efficacy, and safety of a permeation-enhanced testosterone transdermal system in comparison with bi-weekly injections of testosterone enanthate for the treatment of hypogonadal men
The Journal of Clinical Endocrinology and Metabolism, 84(10), 3469-3478 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service