Skip to Content
Merck
All Photos(1)

Key Documents

T0240000

Testosterone decanoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17β-Hydroxy-4-androsten-3-one 17-decanoate, 4-Androsten-17β-ol-3-one 17-caprate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H46O3
CAS Number:
Molecular Weight:
442.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

testosterone

manufacturer/tradename

EDQM

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C29H46O3/c1-4-5-6-7-8-9-10-11-27(31)32-26-15-14-24-23-13-12-21-20-22(30)16-18-28(21,2)25(23)17-19-29(24,26)3/h20,23-26H,4-19H2,1-3H3/t23-,24-,25-,26-,28-,29-/m0/s1

InChI key

LBERVHLCXUMDOT-MPZZESAYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone decanoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Haidl et al.
Der Urologe. Ausg. A, 47(12), 1555-1556 (2008-11-18)
Evidence-based drug therapy for male infertility is often difficult because 30% of all cases of male infertility are classified as idiopathic, and another 30% need surgical treatment. Without knowledge of the underlying pathology, there is no foundation for a specific
Determination of testosterone, testosterone propionate, testosterone enanthate, testosterone undecanoate in pharmaceutical preparations by HPLC.
S Claussen et al.
Die Pharmazie, 55(2), 147-148 (2000-03-21)
J C do Prado et al.
Parasitology research, 85(10), 826-829 (1999-09-24)
Effects of orchiectomy on male Calomys callosus infected with the Y strain of Trypanosoma cruzi were studied. Male C. callosus of the same age and weight were divided into three groups: intact, sham operated, and castrated. After 1 month they
R W Hamill et al.
Brain research, 299(2), 331-337 (1984-05-14)
The effects of the hormone testosterone on neurotransmitter synthesis in peripheral sympathetic ganglia were examined in adult male Sprague-Dawley rats. Tyrosine hydroxylase (T-OH), the rate limiting enzyme in catecholamine biosynthesis was examined in the hypogastric (HG), coeliac (CG), and superior
H A Verheul et al.
Immunopharmacology, 11(2), 93-99 (1986-04-01)
Nandrolone decanoate injections (20, 40 or 80 mg/kg/3 weeks) in female or castrated male New Zealand black/white mice, starting at various ages (4, 9, 17 or 26 weeks) prolong survival, reduce proteinuria and affect the weights of various endocrine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service