Skip to Content
Merck
All Photos(1)

Key Documents

T0250000

Testosterone enantate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Testosterone enanthate, (17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 17β-Hydroxy-4-androsten-3-one 17-enanthate, 4-Androsten-17β-ol-3-one 17-enanthate, NSC 17591, Testosterone 17β-heptanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

testosterone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1

InChI key

VOCBWIIFXDYGNZ-IXKNJLPQSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone enantate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Carole A Samango-Sprouse et al.
American journal of medical genetics. Part A, 161A(3), 501-508 (2013-01-25)
The effects of early androgen treatment on neurodevelopmental performance in pre-pubertal boys with 47,XXY have not been well investigated. The influence of hormones on brain development in humans suggests that a positive effect on neurodevelopmental outcome in young boys with
A F Radicioni et al.
Clinical endocrinology, 76(1), 72-77 (2011-07-02)
Hypogonadism in Prader-Willi syndrome (PWS) is generally attributed to hypothalamic dysfunction or to primary gonadal defect, but pathophysiology is still unclear. To investigate the aetiology of hypothalamic-pituitary-gonadal axis dysfunction in PWS males. Clinical examination and blood sampling for luteinizing hormone
Shane Rogerson et al.
Journal of strength and conditioning research, 21(2), 354-361 (2007-05-29)
Use of testosterone enanthate has been shown to significantly increase strength within 6-12 weeks of administration (2, 9), however, it is unclear if the ergogenic benefits are evident in less than 6 weeks. Testosterone enanthate is classified as a prohibited
Akira Tsujimura et al.
Asian journal of andrology, 13(4), 558-562 (2011-04-05)
Androgens, the levels of which decrease with ageing, play many physiological roles in various organs. Testosterone deficiency syndrome (TDS) has received widespread attention in the last several years. First-line treatment for TDS should be testosterone replacement therapy (TRT), which is
Luigi Di Luigi et al.
The journal of sexual medicine, 9(3), 873-886 (2012-02-11)
A well-tailored testosterone replacement treatment (TRT) in male hypogonadal athletes plays a pivotal role to restore physiological performances, to reduce health risks, and to guarantee the ethic of competition. Few studies evaluated individual androgens profiles during TRT in trained individuals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service