Skip to Content
Merck
All Photos(1)

Key Documents

SML3076

Sigma-Aldrich

Pazopanib hydrochloride

≥98% (HPLC)

Synonym(s):

5-[[4-[(2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl]amino]-2-methylbenzenesulfonamide hydrochloride, 5-[[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]-2-pyrimidinyl]amino]-2-methylbenzenesulfonamide, hydrochloride, GW 786034 HCl, GW 786034B, GW786034 HCl, GW786034B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N7O2S · xHCl
CAS Number:
Molecular Weight:
437.52 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Pazopanib is an orally active receptor tyrosine kinases inhibitor against VEGFR1/2/3 (IC50 = 10/30/47 nM), PDGFR1/2 (IC50 = 84/71 nM) and c-Kit (IC50 = 74 nM), exhibiting reduced or no potency toward FGFR1/3 (IC50 = 140/130 nM), c-fms (IC50 = 146 nM), and other kinases tested (IC50 from >400 nM to >20 μM). Pazopanib exhibits anti-cancer efficacy in cultures (IC50 from 5 to 15 μg/mL; multiple myeloma migration and survival) and in mice in vivo (30-100 mg/kg/day p.o. against human MM xenografts) via angiogenesis inhibition and apoptosis induction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katerina Stamati et al.
Journal of tissue engineering, 11, 2041731420920597-2041731420920597 (2020-06-04)
Pazopanib is a tyrosine kinase inhibitor used to treat renal cell carcinoma. Few in vitro studies investigate its effects towards cancer cells or endothelial cells in the presence of cancer. We tested the effect of Pazopanib on renal cell carcinoma
Adrian Georg Simon et al.
Journal of cancer research and clinical oncology, 146(9), 2255-2265 (2020-06-14)
To investigate the synergistic effect of glycolysis inhibition on therapy answer to tyrosine kinase inhibitors in renal carcinoma. Primary cell cultures from 33 renal tumors including clear cell RCC (ccRCC), papillary RCC and the rare subtype chromophobe RCC as well
Hideki Imano et al.
Toxicology in vitro : an international journal published in association with BIBRA, 71, 105063-105063 (2020-12-04)
Vascular endothelial growth factor (VEGF) promotes tumor angiogenesis through stimulating the proliferation and survival of endothelial cells. The severe adverse events caused by VEGF inhibitors might include immune-related ones; however, details of the mechanism have not been elucidated. We tested
Rakesh Kumar et al.
Molecular cancer therapeutics, 6(7), 2012-2021 (2007-07-11)
With the development of targeted therapeutics, especially for small-molecule inhibitors, it is important to understand whether the observed in vivo efficacy correlates with the modulation of desired/intended target in vivo. We have developed a small-molecule inhibitor of all three vascular
Chiao-Yun Lin et al.
Journal of molecular medicine (Berlin, Germany), 98(8), 1175-1188 (2020-07-09)
Pazopanib-a multitargeted tyrosine kinase inhibitor with prominent antiangiogenic effects-has shown promise in the treatment of soft-tissue sarcomas. Hyperthermia has been also applied as an adjunctive treatment to chemotherapy for these malignancies. Here, we show that pazopanib and hyperthermia act synergistically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service