Skip to Content
Merck
All Photos(1)

Documents

PZ0191

Sigma-Aldrich

Crizotinib

≥98% (HPLC)

Synonym(s):

(R)-3-[1-(2,6-Dichloro-3-fluoro-phenyl)-ethoxy]-5-(1-piperidin-4-yl-1H-pyrazol-4-yl)-pyridin-2-ylamine, PF 2341066, PF-02341066, PF02341066, Xalkori

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22Cl2FN5O
CAS Number:
Molecular Weight:
450.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

storage temp.

room temp

SMILES string

C[C@@H](Oc1cc(cnc1N)-c2cnn(c2)C3CCNCC3)c4c(Cl)ccc(F)c4Cl

InChI

1S/C21H22Cl2FN5O/c1-12(19-16(22)2-3-17(24)20(19)23)30-18-8-13(9-27-21(18)25)14-10-28-29(11-14)15-4-6-26-7-5-15/h2-3,8-12,15,26H,4-7H2,1H3,(H2,25,27)/t12-/m1/s1

InChI key

KTEIFNKAUNYNJU-GFCCVEGCSA-N

Gene Information

Application

Crizotinib has been used:
  • to investigate its effects on hepatocyte growth factor receptor (c-Met) expression, proliferation and apoptosis
  • to block neurotrophic tyrosine kinase receptor type 1 (NTRK1) activity in epithelial cells
  • to restore sensitivity to erlotinib

Biochem/physiol Actions

Crizotinib (PF-02341066) is an ATP-competitive inhibitor of the receptor tyrosine kinases (RTKs) c-Met (hepatocyte growth factor receptor) and anaplastic lymphoma kinase (ALK). Crizotinib is a highly specific inhibitor of c-Met and ALK among > 120 different RTKs surveyed. Crizotinib was approved for treatment of a subtype of nonsmall-cell lung cancer (NSCLC) with ALK fusion mutations.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Registered report: Widespread potential for growth factor-driven resistance to anticancer kinase inhibitors
Greenfield E, et al.
eLife, 3, e04037-e04037 (2014)
Zheng Liu et al.
Oncoimmunology, 9(1), 1758003-1758003 (2020-09-15)
Despite some of the oncogenic driver mutations that have been associated with increased expression of programmed death-ligand 1 (PD-L1), the correlation between PD-L1 expression and ROS1 fusion in NSCLC cells, especially for those with Crizotinib resistance has not been fully
Damini Chand et al.
Disease models & mechanisms, 6(2), 373-382 (2012-10-30)
Neuroblastoma is a childhood extracranial solid tumour that is associated with a number of genetic changes. Included in these genetic alterations are mutations in the kinase domain of the anaplastic lymphoma kinase (ALK) receptor tyrosine kinase (RTK), which have been
Neurotrophic tyrosine kinase receptor 1 is a direct transcriptional and epigenetic target of IL-13 involved in allergic inflammation
Rochman M, et al.
Mucosal Immunology, 8(4), 785-785 (2015)
Nicole R Infarinato et al.
Cancer discovery, 6(1), 96-107 (2015-11-12)
Neuroblastomas harboring activating point mutations in anaplastic lymphoma kinase (ALK) are differentially sensitive to the ALK inhibitor crizotinib, with certain mutations conferring intrinsic crizotinib resistance. To overcome this clinical obstacle, our goal was to identify inhibitors with improved potency that

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service