Skip to Content
Merck
All Photos(1)

Key Documents

SML2282

Sigma-Aldrich

Everolimus

≥95% (HPLC), powder, P-glycoprotein substrate

Synonym(s):

Everolimus, 40-O-(2-Hydroxyethyl)rapamycin, 42-O-(2-Hydroxyethyl)-rapamycin, RAD 001, RAD001, SDZ-RAD, 42-O-(2-Hydroxyethyl)rapamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Everolimus, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(C([C@@]1(O)[C@H](C)CC[C@](C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C2=O)([H])O1)=O)N3[C@H](C(O[C@@]([C@@H](C[C@@]4([H])C[C@@H](OC)[C@H](OCCO)CC4)C)([H])CC([C@H](C)/C=C(C)/[C@@H](O)[C@H]2OC)=O)=O)CCCC3

InChI

1S/C53H83NO14/c1-32-16-12-11-13-17-33(2)44(63-8)30-40-21-19-38(7)53(62,68-40)50(59)51(60)54-23-15-14-18-41(54)52(61)67-45(35(4)28-39-20-22-43(66-25-24-55)46(29-39)64-9)31-42(56)34(3)27-37(6)48(58)49(65-10)47(57)36(5)26-32/h11-13,16-17,27,32,34-36,38-41,43-46,48-49,55,58,62H,14-15,18-26,28-31H2,1-10H3/b13-11+,16-12+,33-17+,37-27+/t32-,34-,35-,36-,38-,39+,40+,41+,43-,44+,45+,46-,48-,49+,53-/m1/s1

InChI key

HKVAMNSJSFKALM-GKUWKFKPSA-N

Gene Information

human ... FKBP1A(2280)

Looking for similar products? Visit Product Comparison Guide

Application

Everolimus has been used as an mechanistic target of rapamycin complex 1 (mTORC1) inhibitor to study its effect on MCF7 and MDA-MB-231 cell lines.

Biochem/physiol Actions

Everolimus, a substrate for P-glycoprotein (P-gp) can be used to treat advanced neuroendocrine tumors (NETs).
Everolimus, the 40-O-(2-hydroxyethyl) derivative of rapamycin (sirolimus), is a potent and selective inhibitor of mechanistic target of rapamycin (mTOR). Everolimus is selective for the mTORC1 protein complex. Everolimus exhibit potent immunosuppressive and anticancer activities.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Testes

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical modification of rapamycin: the discovery of SDZ RAD.
R Sedrani et al.
Transplantation proceedings, 30(5), 2192-2194 (1998-09-02)
O-GlcNAc Transferase Inhibition Differentially Affects Breast Cancer Subtypes
Barkovskaya A, et al.
Scientific Reports, 9(1), 5670-5670 (2019)
Everolimus in the treatment of neuroendocrine tumors: efficacy, side-effects, resistance, and factors affecting its place in the treatment sequence
Lee L, et al.
Expert Opinion on Pharmacotherapy, 19(8), 909-928 (2018)
Aisha Naeem et al.
Molecular cancer research : MCR (2021-10-13)
In medulloblastoma, p53 expression has been associated with chemoresistance and radiation resistance and with poor long-term outcomes in the p53-mutated sonic hedgehog, MYC-p53, and p53-positive medulloblastoma subgroups. We previously established a direct role for p53 in supporting drug resistance in
Matteo Morotti et al.
Nature, 629(8011), 426-434 (2024-04-25)
Expansion of antigen-experienced CD8+ T cells is critical for the success of tumour-infiltrating lymphocyte (TIL)-adoptive cell therapy (ACT) in patients with cancer1. Interleukin-2 (IL-2) acts as a key regulator of CD8+ cytotoxic T lymphocyte functions by promoting expansion and cytotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service