Skip to Content
Merck
All Photos(1)

Key Documents

475991

Sigma-Aldrich

Torin1

≥99% (HPLC), powder, mTOR inhibitor, Calbiochem®

Synonym(s):

mTOR Inhibitor XI, Torin1, 1-(4-(4-propionylpiperazin-1-yl)-3-(trifluoromethyl)phenyl)-9-(quinolin-3-yl)benzo[h][1,6]naphthyridin-2(1H)-one, DNA-PK Inhibitor VI, PI 3-K Inhibitor XVIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H28F3N5O2
CAS Number:
Molecular Weight:
607.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

mTOR Inhibitor XI, Torin1, mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).

Quality Level

Assay

≥99% (HPLC)

form

powder

potency

2 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white

solubility

DMSO: 2 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(ccc(c1)[n]3c4c5c(ncc4cc[c]3=O)ccc(c5)c6cnc7c(c6)cccc7)N2CCN(CC2)C(=O)CC

InChI

1S/C35H28F3N5O2/c1-2-32(44)42-15-13-41(14-16-42)31-11-9-26(19-28(31)35(36,37)38)43-33(45)12-8-24-20-40-30-10-7-22(18-27(30)34(24)43)25-17-23-5-3-4-6-29(23)39-21-25/h3-12,17-21H,2,13-16H2,1H3

InChI key

AKCRNFFTGXBONI-UHFFFAOYSA-N

General description

A cell-permeable pyridinonequinoline derivative that acts as a highly potent, ATP-competitive inhibibitor against mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively, in cell-free kinase assays), inhibiting class I (IC50 =171, 250, and 564 nM, respectively, against P110γ, P110α/P85α, and P110δ/P85α, respectively), class II (IC50 =176 and 564 nM, respectively, against C2α, and C2β, respectively), and class III (IC50 = 533 nM against hVPS34) PI 3-K only at much higher concentrations. Effectively inhibits mTORC1-mediated S6K1 Thr389 phosphorylation in MEF cultures (IC50 = 2 nM) in vitro, as well as mTORC2-mediated Akt Ser473 and mTORC1-dependent S6 Ser235/236 phosphorylations in murine lung and liver in vivo (up to 6 h after single i.p. dose at 20 mg/kg). Despite its poor in vivo stability (T1/2 = 4.52 h in mice; 10 mg/kg i.p.), Torin1 is reported to completely suppress U87MG-derived tumor expansion in mice when administered via daily i.p. at a high dosage of 200 mg/kg. Also reported to effectively inhibit cellular mTORC1 functionalities known to be rapamycin-resistant (Cat. Nos. 553210, 553211, and 553212M).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MTOR
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20&degC). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Liu, Q., et al. 2010. J Med Chem.53, 7146.
Thoreen, C., et al. 2009. J Biol Chem.284, 8023.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takafumi Miyamoto et al.
Cell reports methods, 1(4), 100052-100052 (2022-04-28)
Engineered synthetic biomolecular devices that integrate elaborate information processing and precisely regulate living cell behavior have potential in various applications. Although devices that directly regulate key biomolecules constituting inherent biological systems exist, no devices have been developed to control intracellular
Yucel Aydin et al.
Journal of hepatocellular carcinoma, 8, 1579-1596 (2021-12-18)
Hepatocellular carcinoma (HCC) developing in the context of preexisting cirrhosis is characterized by impaired autophagy that results in increased exosome release. This study was conducted to determine whether circulating exosomes expressing glypican 3 (GPC3) could be utilized as a biomarker
Madison C Williams et al.
Developmental biology, 473, 59-70 (2021-01-24)
Xenopus tadpoles are a unique model for regeneration in that they exhibit two distinct phases of age-specific regenerative competence. In Xenopus laevis, young tadpoles fully regenerate following major injuries such as tail transection, then transiently lose regenerative competence during the
Jee-Yun Park et al.
Scientific reports, 11(1), 21119-21119 (2021-10-28)
Transcription factor EB (TFEB) is a master regulator of the autophagy-lysosomal pathway (ALP). Here, we cloned a novel splicing variant of TFEB, comprising 281 amino acids (hereafter referred to as small TFEB), and lacking the helix-loop-helix (HLH) and leucine zipper
Manasa P Srikanth et al.
Stem cells translational medicine, 10(7), 1081-1094 (2021-03-04)
Gaucher disease (GD) is a lysosomal storage disorder caused by mutations in GBA1, the gene that encodes lysosomal β-glucocerebrosidase (GCase). Mild mutations in GBA1 cause type 1 non-neuronopathic GD, whereas severe mutations cause types 2 and 3 neuronopathic GD (nGD). GCase deficiency

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service