Skip to Content
Merck
All Photos(1)

Key Documents

SML2263

Sigma-Aldrich

Ladostigil tartrate

≥98% (HPLC)

Synonym(s):

(3R)-(N-Propargylaminoindan-5-yl)-ethyl methyl carbamate tartrate (2:1), TV-3326, TV3326

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O2 · 0.5C4H6O6
CAS Number:
Molecular Weight:
347.39
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/2C16H20N2O2.C4H6O6/c2*1-4-10-17-15-9-7-12-6-8-13(11-14(12)15)20-16(19)18(3)5-2;5-1(3(7)8)2(6)4(9)10/h2*1,6,8,11,15,17H,5,7,9-10H2,2-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t2*15-;1-,2-/m111/s1

InChI key

PRLVBVAJMQZMJN-OGOSNNLPSA-N

Biochem/physiol Actions

Ladostigil, a carbamate is also a butyryl cholinesterase (BuChE) inhibitor. It possesses anti-inflammatory property and effectively reduces the levels of tumor necrosis factor α (TNF-α) in lipopolysaccharide (LPS)-activated macrophages. It provides protection to cardiomyocytes. Ladostigil has neuroprotective functionality and has the potential to treat mild cognitive impairment (MCI) and spatial memory deficits development. It also exhibits protective effects during oxidative and nitrative stress.
Multifunctional drug designed to treat Alzhemier′s disease by combining in a single molecule the neuroprotective/neurorestorative effects of the (MAO)-B inhibitor rasagiline with the cholinesterase (ChE) inhibitory activity of rivastigmine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moussa B H Youdim
Experimental neurobiology, 22(1), 1-10 (2013-04-16)
Present anti-PD and -AD drugs have limited symptomatic activity and devoid of neuroprotective and neurorestorative property that is needed for disease modifying action. The complex pathology of PD and AD led us to develop several multi-target neuroprotective and neurorestorative drugs
Rony Panarsky et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 7(2), 488-498 (2012-03-29)
Impaired mitochondrial function accompanied by microglial activation and the release of nitric oxide (NO) and pro-inflammatory cytokines has been reported in Alzheimer's disease, its prodromal phase of Mild Cognitive Impairment (MCI) and in aged rats. The present study showed that
Inessa Yanovsky et al.
Journal of medicinal chemistry, 55(23), 10700-10715 (2012-11-16)
The cascade of events that occurs in Alzheimer's disease involving oxidative stress and the reduction in cholinergic transmission can be better addressed by multifunctional drugs than cholinesterase inhibitors alone. For this purpose, we prepared a large number of derivatives of
Orly Weinreb et al.
British journal of pharmacology, 173(13), 2080-2094 (2015-09-04)
Alzheimer's disease (AD) is accepted nowadays as a complex neurodegenerative disorder with multifaceted cerebral pathologies, including extracellular deposition of amyloid β peptide-containing plaques, intracellular neurofibrillary tangles, progressive loss of cholinergic neurons, metal dyshomeostasis, mitochondrial dysfunction, neuroinflammation, glutamate excitoxicity, oxidative stress
John P M Finberg et al.
Frontiers in pharmacology, 7, 340-340 (2016-11-03)
Inhibitors of MAO-A and MAO-B are in clinical use for the treatment of psychiatric and neurological disorders respectively. Elucidation of the molecular structure of the active sites of the enzymes has enabled a precise determination of the way in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service