Skip to Content
Merck
All Photos(1)

Key Documents

SML2012

Sigma-Aldrich

Cinacalcet hydrochloride

≥98% (HPLC)

Synonym(s):

AMG 073 hydrochloride, KRN 1493 hydrochloride, N-[1-(R)-(-)-(1-naphthyl)ethyl]-3-[3(trifluoromethyl)phenyl]-1-aminopropane hydrochloride, NPS 1493 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22F3N·HCl
CAS Number:
Molecular Weight:
393.87
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -24 to -32°, c = 0.5 in methanol

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/t16-;/m1./s1

InChI key

QANQWUQOEJZMLL-PKLMIRHRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cinacalcet hydrochloride has been used:
  • to stimulate calcium-sensing receptor (CaSR) in thick ascending limb (TAL) of the male Sprague-Dawley rats to study the effect of altered claudins levels on calciuria and magnesiuria in metabolic acidosis
  • as a calcimimetic positive allosteric modulator (PAM) of CaSR to determine the structural rearrangements of human CaSR upon activation
  • as a calcium-sensing receptor (CaSR) agonist to study the effect of CaSR on tumor cell proliferation in lung adenocarcinoma (LUAD) cell line

Biochem/physiol Actions

Cinacalcet is a positive allosteric modulator of the Calcium Sensing Receptor (CaSR) that acts as a calcimimetic. It increases the sensitivity of the calcium-sensing receptor to extracellular calcium resulting in a decrease in serum calcium levels.
Studies show that oral administration of cinacalcet hydrochloride aids in treating secondary hyperparathyroidism in patients with chronic kidney disease on dialysis. It may also be a potential therapeutic to treat hypercalcemia in patients with parathyroid carcinoma.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1B - STOT RE 2

Target Organs

eye - retina

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yang Gao et al.
Nature, 595(7867), 455-459 (2021-07-02)
The calcium-sensing receptor (CaSR), a cell-surface sensor for Ca2+, is the master regulator of calcium homeostasis in humans and is the target of calcimimetic drugs for the treatment of parathyroid disorders1. CaSR is a family C G-protein-coupled receptor2 that functions
Jordi Bover et al.
Clinical journal of the American Society of Nephrology : CJASN, 11(1), 161-174 (2015-08-01)
CKD and CKD-related mineral and bone disorders (CKD-MBDs) are associated with high cardiovascular and mortality risks. In randomized clinical trials (RCTs), no single drug intervention has been shown to reduce the high mortality risk in dialysis patients, but several robust
Jiansha Li et al.
Frontiers in cell and developmental biology, 8, 446-446 (2020-07-17)
The effect of calcium sensing receptor (CaSR) on tumor cell proliferation has been studied in several human cancers, and great discrepancies were found in different tumors. However, the role of CaSR in lung adenocarcinomas (LUADs) is not clear. Therefore, we
Gamal A E Mostafa et al.
Profiles of drug substances, excipients, and related methodology, 42, 1-90 (2017-04-23)
Cinacalcet hydrochloride is a calcimimetic agent that increases the sensitivity to the extracellular calcium of the calcium-sensing receptors of the parathyroid gland which regulates parathyroid hormone secretion. This comprehensive profile on cinacalcet hydrochloride starts with a description: nomenclature, formulae, chemical
Il Hwan Oh et al.
American journal of physiology. Renal physiology, 320(3), F418-F428 (2021-02-02)
Urinary calcium and magnesium wasting is a characteristic feature of metabolic acidosis, and this study focused on the role of the thick ascending limb of Henle's loop in metabolic acidosis-induced hypercalciuria and hypermagnesiuria because thick ascending limb is an important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service