Skip to Content
Merck
All Photos(2)

Key Documents

L7757

Sigma-Aldrich

Lidocaine

≥98% (GC), powder, neuroprotective agent

Synonym(s):

2-Diethylamino-N-(2,6-dimethylphenyl)acetamide, Lignocaine, Xylocaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Lidocaine, powder

form

powder

SMILES string

CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)

InChI key

NNJVILVZKWQKPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lidocaine is a local anesthetic and an effective antiarrhythmia agent. It occurs as solutions of crystalline colorless solid for injections, that has a short half-life within plasma.

Application

Lidocaine may be used for drug testing and for voltage clamp experiments.

Biochem/physiol Actions

Lidocaine, a neuroprotective agent, mediates blockage of sodium (Na+) channels. It prevents the influx of sodium ions leading to depolarization of sodium channels and metabolically inactivated in liver. Lidocaine has positive effect on cerebral ischemia. It is also used in the treatment of ventricular tachycardia. Lidocaine is an efficient anesthetic agent in ophthalmic surgeries as it is highly potent and has good tissue penetrability.
Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Na+ channel blocker; class IB antiarrhythmic.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetics and pharmacodynamics of lignocaine: a review
Weinberg L, et al.
World Journal of Anesthesiology, 4(2), 17-29 (2015)
Mitochondrial injury and caspase activation by the local anesthetic lidocaine
Johnson ME, et al.
Anesthesiology, 101(5), 1184-1194 (2004)
A microfluidic gradient maker for toxicity testing of bupivacaine and lidocaine
Tirella A, et al.
Toxicology in vitro, 22(8), 1957-1964 (2008)
Neuroprotective effect of low-dose lidocaine in a rat model of transient focal cerebral ischemia
Lei B, et al.
Anesthesiology, 95(2), 445-451 (2001)
Principles of Pharmacology: The Pathophysiologic Basis of Drug Therapy (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service