Skip to Content
Merck

Skip To

SML0650

Risedronate sodium

≥97% (HPLC)

Synonym(s):

P,P′-[1-Hydroxy-2-(3-pyridinyl)ethylidene]bis-phosphonic acid sodium, Sodium risedronate, [1-Hydroxy-2-(3-pyridinyl)ethylidene]bis[phosphonic acid] monosodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H10NO7P2 · Na
CAS Number:
Molecular Weight:
305.09
UNSPSC Code:
41106300
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

[Na+].[P](=O)([O-])(O)[C@]([P](=O)(O)O)(O)Cc1cnccc1

InChI

1S/C7H11NO7P2.Na/c9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6;/h1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15);/q;+1/p-1

InChI key

DRFDPXKCEWYIAW-UHFFFAOYSA-M

Biochem/physiol Actions

Risedronate sodium is a bisphosphonate bone resorption inhibitor.
Risedronate sodium is a bisphosphonate bone resorption inhibitor. It has an affinity for hydroxyapatite crystals in bone and acts as an antiresorptive agent and is an inhibitor of farnesyl diphosphate (FPP) synthase, which results in downstream inhibition of osteoclast activity and reduced bone resorption and turnover. Risedronate sodium has been used to treat postmenopausal osteoporosis and Paget′s disease.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A4978F86821604632
description

≥97% (HPLC)

description

≥97% (NMR), powder

description

≥95% (NMR)

description

United States Pharmacopeia (USP) Reference Standard

assay

≥97% (HPLC)

assay

≥97% (NMR)

assay

≥95% (NMR)

assay

-

form

powder

form

powder

form

powder

form

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

room temp

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

solubility

H2O: 5 mg/mL, clear (warmed)

solubility

water, double-distilled: 10 mg/mL

solubility

H2O: 20 mg/mL, clear

solubility

-

color

white to beige

color

white

color

white to beige

color

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters