Skip to Content
Merck
All Photos(1)

Documents

A4978

Sigma-Aldrich

Alendronate sodium trihydrate

≥97% (NMR), powder

Synonym(s):

4-amino-1-hydroxy-1-phosphonobutyl phosphonic acid, monosodium, MK-217

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12NaNO7P2 · 3 H2O
CAS Number:
Molecular Weight:
325.12
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (NMR)

form

powder

color

white

solubility

water, double-distilled: 10 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].NCCCC(O)(P(O)(O)=O)P(O)([O-])=O

InChI

1S/C4H13NO7P2.Na.3H2O/c5-3-1-2-4(6,13(7,8)9)14(10,11)12;;;;/h6H,1-3,5H2,(H2,7,8,9)(H2,10,11,12);;3*1H2/q;+1;;;/p-1

InChI key

DCSBSVSZJRSITC-UHFFFAOYSA-M

Gene Information

human ... FDPS(2224)

Looking for similar products? Visit Product Comparison Guide

Application

Alendronate sodium trihydrate has been used:
  • for calcium phosphate nanoparticles (nCaP)/ bisphosphonate (BP) coating on titanium implants and analyze its osteogenic capacity
  • in osteogenic differentiation of human bone marrow stromal cells (hBMSCs) isolated from aged osteoporosis patients- and healthy-person
  • to treat osteoblast progenitor cells and osteoblasts and study their effect on osteoblast behaviors using conventional assays and simple digital image analysis

Biochem/physiol Actions

Alendronate is a nitrogen-containing bisphosphonate that is known to hinder the mevalonate pathway and limit osteoclastic function and activity. Alendronate sodium trihydrate is a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor, and CD45 protein tyrosine phosphatase inhibitor.
Alendronate sodium trihydrate is a bone resorption inhibitor; farnesyl diphosphate synthase inhibitor (IC50 = 460 nM); CD45 protein tyrosine phosphatase inhibitor.

Storage and Stability

Following reconstitution, it is recommended to aliquot and freeze at -20°C. Stock solutions are stable for up to 6 months at -20°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shmuel Banai et al.
American heart journal, 165(2), 234-240 (2013-01-29)
Activation of systemic innate immunity is critical in the chain of events leading to restenosis. LABR-312 is a novel compound that transiently modulates circulating monocytes, reducing accumulation of these cells at vascular injury sites and around stent struts. The purpose
Xuefeng Hu et al.
Tissue engineering. Part A, 19(17-18), 1919-1930 (2013-04-02)
Fibrous encapsulation can impair implant osseointegration and cause implant failure but currently there are limited strategies to address this problem. Since bisphosphonates (BPs), a class of drugs widely used to treat bone diseases, was recently found to induce fibroblast apoptosis
Oh Sasaki et al.
Journal of immunology (Baltimore, Md. : 1950), 191(6), 2879-2889 (2013-08-13)
Bisphosphonates (BPs) have been widely used to treat osteoporosis. They act by inhibiting farnesyl diphosphate synthase in the mevalonate pathway. This resembles the action of statins, whose immune-modulating effect has recently been highlighted. In contrast, the effect of BPs on
Young-Pil Yun et al.
Journal of biomedical nanotechnology, 10(6), 1080-1090 (2014-04-23)
The osteogenic effect of culturing adipose-derived stem cells (ADSCs) on alendronate (Aln)-loaded polycarprolactone (PCL) nanofibrous scaffolds was evaluated by examining alkaline phosphatase (ALP) activity, calcium content, and expression of osteogenic differentiation genes in vitro. The 10% Aln/PCL nanofibrous scaffolds showed
Núria Guañabens et al.
Hepatology (Baltimore, Md.), 58(6), 2070-2078 (2013-05-21)
Osteoporosis resulting in bone fractures is a complication in patients with primary biliary cirrhosis (PBC). Once-weekly alendronate improves bone mass and is well tolerated in these patients, but there is a concern because of poor compliance. Therefore, the efficacy, adherence

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service