Skip to Content
Merck

Skip To

1604632

USP

Risedronate Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,6-Bis[(3-pyridinyl)methyl]-2,5-dihydroxy-2,5-dioxido-1,4,2,5-dioxadiphosphorinane-3,6-diyl]bis[phosphonic acid] disodium salt tetrahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H16N2Na2O12P4 · 4H2O
Molecular Weight:
646.22
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

risedronate

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

OP1(OC(CC2=CN=CC=C2)(P([O-])(O)=O)P(OC1(CC3=CC=CN=C3)P([O-])(O)=O)(O)=O)=O.O.O.O.O.[Na+].[Na+]

InChI

1S/C14H18N2O12P4.2Na.4H2O/c17-29(18,19)13(7-11-3-1-5-15-9-11)27-32(25,26)14(30(20,21)22,28-31(13,23)24)8-12-4-2-6-16-10-12;;;;;;/h1-6,9-10H,7-8H2,(H,23,24)(H,25,26)(H2,17,18,19)(H2,20,21,22);;;4*1H2/q;2*+1;;;;/p-2

InChI key

JBPZPQPSFCJGDM-UHFFFAOYSA-L

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
160462116046431598609
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

agency

USP

agency

USP

agency

USP

agency

USP

API family

risedronate

API family

risedronate

API family

risedronate

API family

ranitidine


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters