Skip to Content
Merck

Skip To

68264

α-Hydroxyisobutyronitrile β-D-glucopyranoside

≥97% (HPLC)

Synonym(s):

α-Hydroxyisobutyronitrile β-D-glucose, 2-(β-D-Glucopyranosyloxy)-2-methylpropionitrile, Linamarin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H17NO6
CAS Number:
Molecular Weight:
247.25
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥97% (HPLC)

form

solid

optical activity

[α]/D -26.5±2.0°, c = 1 in H2O

technique(s)

HPLC: suitable

color

white to off-white

storage temp.

2-8°C

SMILES string

CC(C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)C#N

InChI

1S/C10H17NO6/c1-10(2,4-11)17-9-8(15)7(14)6(13)5(3-12)16-9/h5-9,12-15H,3H2,1-2H3/t5-,6-,7+,8-,9+/m1/s1

InChI key

QLTCHMYAEJEXBT-ZEBDFXRSSA-N

Application

Linamarin, a cyanogenic glucose substrate, is used together with β-glucosidase, linamarase, to produce cyanide in vivo as a potential anticancer strategy.

Biochem/physiol Actions

Linamarin is a cyanogenic glucoside found in the leaves and roots of plants such as cassava, lima beans, and flax. Upon exposure to enzymes and gut flora in the human intestine, linamarin and its methylated relative lotaustralin can decompose to the toxic chemical hydrogen cyanide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10050M0779G9662
description

≥97% (HPLC)

description

BioXtra, ≥97.0% (HPLC)

description

≥99% (HPLC and GC)

description

≥90%

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

biological source

synthetic

biological source

synthetic

biological source

-

biological source

synthetic

assay

≥97% (HPLC)

assay

≥97.0% (HPLC)

assay

≥99% (HPLC and GC)

assay

≥90%

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

solid

form

powder

form

powder

form

powder or solid

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters