Skip to Content
Merck
All Photos(1)

Key Documents

07743

Sigma-Aldrich

Dhurrin

≥95% (HPLC)

Synonym(s):

(S)-(β-D-Glucopyranosyloxy)(4-hydroxyphenyl)acetonitrile, (S)-4-Hydroxymandelonitrile β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO7
CAS Number:
Molecular Weight:
311.29
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder or crystals

color

white to light brown

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H](O[C@H](C#N)c2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O
OC[C@H]1O[C@@H](O[C@H](C#N)c2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H17NO7/c15-5-9(7-1-3-8(17)4-2-7)21-14-13(20)12(19)11(18)10(6-16)22-14/h1-4,9-14,16-20H,6H2/t9-,10-,11-,12+,13-,14-/m1/s1

InChI key

NVLTYOJHPBMILU-YOVYLDAJSA-N

General description

Dhurrin is a cyanogenic glucoside found expressed in the leaves of Sorghum plants. Studies have shown that as the plant ages, dhurrin expression depends are nitrogen availability. Dhurrin production shifts from the leaves to the stems during development. [1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang Chul Choi et al.
Plants (Basel, Switzerland), 9(10) (2020-10-23)
Sorghum (Sorghum bicolor (L.)) Moench is an important food for humans and feed for livestock. Sorghum contains dhurrin which can be degraded into toxic hydrogen cyanide. Here, we report the expression patterns of 14 candidate genes related to dhurrin ((S)-4-Hydroxymandelnitrile-β-D-glucopyranoside)
Plant science. Dynamic metabolons.
Birger Lindberg Møller
Science (New York, N.Y.), 330(6009), 1328-1329 (2010-12-04)
Kirsten A Nielsen et al.
Planta, 223(5), 1010-1023 (2005-11-25)
Barley (Hordeum vulgare L.) produces a leucine-derived cyanogenic beta-D-glucoside, epiheterodendrin that accumulates specifically in leaf epidermis. Barley leaves are not cyanogenic, i.e. they do not possess the ability to release hydrogen cyanide, because they lack a cyanide releasing beta-D-glucosidase. Cyanogenesis
Cyanogenic glycosides and menisdaurin from Guazuma ulmifolia, Ostrya virgininana, Tiquilia plicata and Tiquilia canescens.
David S Seigler
Phytochemistry, 66(13), 1567-1580 (2005-07-09)
Gina Rosalinda De Nicola et al.
Journal of agricultural and food chemistry, 59(15), 8065-8069 (2011-06-29)
Cyanogenic plants have some potential as biocidal green manure crops in limiting several soilborne pests and pathogens. Sorghum (Sorghum bicolor (L.) Moench) and Sudangrass (Sorghum bicolor subsp. sudanense (P.) Stapf), in fact, contain the cyanogenic glucoside p-hydroxy-(S)-mandelonitrile-β-D-glucoside (dhurrin) as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service