Skip to Content
Merck
All Photos(1)

Documents

SAB4700587

Sigma-Aldrich

Monoclonal Anti-CD45 antibody produced in rat

clone EM-05, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Ptprc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EM-05, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

General description

The antibody EM-05 reacts with mouse CD45 antigen (Leukocyte Common Antigen), a single chain type I transmembrane protein expressed at high level on cells of hematopoietic origin, except erythrocytes and platelets.

Immunogen

Murine peripheral blood leukocytes

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey E Hespe et al.
Plastic and reconstructive surgery, 143(1), 77e-87e (2018-12-28)
Genetic mutations and obesity increase the risk of secondary lymphedema, suggesting that impaired lymphatic function before surgical injury may contribute to disease pathophysiology. Previous studies show that obesity not only decreases lymphatic function, but also markedly increases pathologic changes, such
Hui Gong et al.
Cellular and molecular life sciences : CMLS, 81(1), 39-39 (2024-01-12)
Colorectal cancer (CRC) is characterized by a complex tumor inflammatory microenvironment, while angiogenesis and immunosuppression frequently occur concomitantly. However, the exact mechanism that controls angiogenesis and immunosuppression in CRC microenvironment remains unclear. Herein, we found that expression levels of lipid
Yuxuan Miao et al.
Cell, 177(5), 1172-1186 (2019-04-30)
Our bodies are equipped with powerful immune surveillance to clear cancerous cells as they emerge. How tumor-initiating stem cells (tSCs) that form and propagate cancers equip themselves to overcome this barrier remains poorly understood. To tackle this problem, we designed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service