Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502541

Sigma-Aldrich

Anti-CD45 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

L-CA, LY-5, Leukocyte common antigen precursor, Leukocyte common antigen variant 4 precursor, Lymphocyte common antigen Ly-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 147 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)

General description

Anti-CD45 antibody detects endogenous levels of total CD45 protein.
The CD45 (cluster of differentiation 45) gene is mapped to human chromosome 1q31.3-q32.1. It codes for a type I transmembrane protein and is expressed in nucleated hematopoietic cells along within its precursors. The protein consists of a varying extracellular domain and a highly conserved intracellular domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD45.

Immunogen Range: 981-1030

Application

Anti-CD45 antibody produced in rabbit has been used in:
  • immunohistochemistry staining[1]
  • immunofluorescence staining[2]
  • immunostaining

Biochem/physiol Actions

The Cluster of differentiation 45 (CD45) facilitates the regulation of lymphocyte signaling and SRC family kinase Lck. This protein participates in the polymorphism in several mammalian species.
The protein encoded by CD45 (cluster of differentiation 45) is a prototypic receptor and exhibits phosphatase activity. CD45 is found to control T-cell mediated BCR-signaling.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sathish Babu Vasamsetti et al.
Immunity, 49(1), 93-106 (2018-07-01)
There is a growing body of research on the neural control of immunity and inflammation. However, it is not known whether the nervous system can regulate the production of inflammatory myeloid cells from hematopoietic progenitor cells in disease conditions. Myeloid
The soluble cytoplasmic tail of CD45 (ct-CD45) in human plasma contributes to keep T cells in a quiescent state.
Puck A
European Journal of Immunology, 47(1), 193-205 (2017)
T-helper signals restore B-cell receptor signaling in autoreactive anergic B cells by upregulating CD45 phosphatase activity.
Szodoray P
The Journal of Allergy and Clinical Immunology, 138(3), 839-851 (2016)
Milja Arponen et al.
Frontiers in physiology, 13, 1035516-1035516 (2022-12-17)
Bone is an active tissue that undergoes constant remodeling. Bone formation requires energy and one of the energy sources of bone-forming osteoblasts is glucose, which is transported inside the cells via glucose transporters. However, the role of class I glucose
Ondřej Kodet et al.
International journal of molecular sciences, 22(13) (2021-07-21)
Therapy targeting immune checkpoints represents an integral part of the treatment for patients suffering from advanced melanoma. However, the mechanisms of resistance are responsible for a lower therapeutic outcome than expected. Concerning melanoma, insufficient stimulation of the immune system by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service