Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500974

Sigma-Aldrich

Anti-Fibronectin Antibody

rabbit polyclonal

Synonym(s):

CIG, FINC, cold-insoluble globulin, fibronectin, fibronectin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Fibronectin 1 antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 262 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)

General description

Anti-Fibronectin 1 antibody detects endogenous levels of total Fibronectin 1 protein.
Fibronectin 1 is a glycoprotein of the extracellular matrix that is coded by FN1 gene. It is expressed in the plasma and at the cell surface. It is mapped to human chromosome 2q35.

Immunogen

The antiserum was produced against synthesized peptide derived from human Fibronectin 1.

Immunogen Range: 2337-2386

Application

Anti-Fibronectin 1, C-Terminal antibody has been used in western blotting, cell staining and immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Fibronectin participates in cell adhesion, growth, migration, wound healing, blood coagulation and metastasis. Mutations in FN1 results in glomerulopathy. It plays an important role in cell attachment and spreading, control of cell cytoskeleton, morphology and differentiation. FN1 is also involved in extracellular matrix formation, hemostasis and thrombosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kusuma Suphakhong et al.
The Journal of biological chemistry, 298(11), 102554-102554 (2022-10-03)
N6-methyladenosine (m6A) is the most common internal chemical modification of mRNAs involved in many pathological processes including various cancers. In this study, we investigated the m6A-dependent regulation of JUN and JUNB transcription factors (TFs) during transforming growth factor-beta-induced epithelial-mesenchymal transition
Minoru Terashima et al.
The Journal of biological chemistry, 293(47), 18016-18030 (2018-09-29)
Long noncoding RNAs (lncRNAs) are important regulatory molecules in various biological and pathological processes, including cancer development. We have previously shown that the MEG3 lncRNA plays an essential role in transforming growth factor-β (TGF-β)-induced epithelial-mesenchymal transition (EMT) of human lung
Jiangtao Yu et al.
Journal of experimental & clinical cancer research : CR, 37(1), 222-222 (2018-09-12)
Emerging evidence has shown long noncoding RNAs (lncRNAs) exert important roles in colorectal cancer (CRC) tumorigenesis. However, most lncRNAs involved in this process remain undefined and the underlying molecular mechanisms mediated by lncRNAs are largely unknown. An unbiased screening was
Colorectal Cancer Aggressiveness is Related to Fibronectin Over Expression, Driving the Activation of SDF-1:CXC
Fernandes SG, et al.
International Journal of Cancer and Clinical Research, 3(8), 72-81 (2016)
Epigenetic regulation of epithelial-mesenchymal transition by KDM6A histone demethylase in lung cancer cells
Terashima M, et al.
Biochemical and Biophysical Research Communications, 490(4), 1407-1413 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service