Skip to Content
Merck
All Photos(1)

Documents

MAB1940

Sigma-Aldrich

Anti-Fibronectin Antibody, cellular, clone DH1

clone DH1, Chemicon®, from mouse

Synonym(s):

Anti-CIG, Anti-ED-B, Anti-FINC, Anti-FN, Anti-FNZ, Anti-GFND, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-SMDCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DH1, monoclonal

species reactivity

rat, guinea pig, human, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)
rabbit ... Fn1(100328589)
rat ... Fn1(25661)

Specificity

Specific for the extra domain of cellular fibronectin (EDA sequence) and thus recognizes only the cellular fibronectin.

Immunogen

Epitope: cellular
Purified fibronectin from Human A8387 fibrosarcoma cells.

Application

Detect Fibronectin using this Anti-Fibronectin Antibody, cellular, clone DH1 validated for use in WB, IH.
Immunoblotting at 1:1,000

Indirect immunohistochemistry at 1:200-1:400

A 1-hour fixation with 3-3.5% paraformaldehyde in 0.1 M phosphate buffer pH 7.4 is suggested. Shows some reactivity on paraffin sections depending on fixation conditions and time (Latvala et al., 1996).

Final working dilutions must be determined by end user.

Physical form

Format: Purified
In PBS with 1% BSA and 0.1% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired elastogenesis in Hurler disease: dermatan sulfate accumulation linked to deficiency in elastin-binding protein and elastic fiber assembly.
Hinek, A; Wilson, SE
The American Journal of Pathology null
Jassim M Al-Hassan et al.
Frontiers in pharmacology, 11, 899-899 (2020-07-07)
Preparations from Arabian Gulf catfish (Arius bilineatus, Val) epidermal gel secretion (PCEGS) effectively heal chronic wounds in diabetic patients. However, specific lipid components of PCEGS that are responsible for various aspects of wound healing are unknown. Here, we report for
Olaf Voets et al.
PloS one, 12(8), e0182974-e0182974 (2017-08-12)
Phenotypic assays using human primary cells are highly valuable tools for target discovery and validation in drug discovery. Expression knockdown (KD) of such targets in these assays allows the investigation of their role in models of disease processes. Therefore, efficient
Grissel Faura Tellez et al.
American journal of physiology. Lung cellular and molecular physiology, 309(7), L725-L735 (2015-07-26)
Genetic studies have identified Protocadherin-1 (PCDH1) and Mothers against decapentaplegic homolog-3 (SMAD3) as susceptibility genes for asthma. PCDH1 is expressed in bronchial epithelial cells and has been found to interact with SMAD3 in yeast two-hybrid (Y2H) overexpression assays. Here, we
Katja Vehanen et al.
Acta ophthalmologica, 86(1), 91-96 (2007-08-23)
The use of biopolymers in peribulbar injection for controlled drug delivery provides alternative options to eyedrops and intravitreal or surgical methods. Polymerizable biopolymers are especially likely to have a role because of their particular properties. In liquid form, they can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service