Skip to Content
Merck
All Photos(2)

Documents

AB1954

Sigma-Aldrich

Anti-Fibronectin Antibody

Chemicon®, from rabbit

Synonym(s):

Cold-insoluble globulin, fibronectin 1, migration stimulating factor, migration-stimulating factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)
mouse ... Fn1(14268)

General description

Fibronectin (FN) is involved in many cellular processes, including tissue repair, embryogenesis, blood clotting, and cell migration/adhesion. Fibronectin exists in two main forms: 1) as an insoluble glycoprotein dimer that serves as a linker in the ECM (extracellular matrix), and; 2) as a soluble disulphide linked dimer found in the plasma (plasma FN). The plasma form is synthesized by hepatocytes, and the ECM form is made by fibroblasts, chondrocytes, endothelial cells, macrophages, as well as certain epithelial cells.

Specificity

Other species have not been tested.
The antibody reacts with the 262 kDa fibronectin protein.

Immunogen

Purified rat plasma fibronectin.

Application

Anti-Fibronectin Antibody detects level of Fibronectin & has been published & validated for use in ELISA & WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Routinely evaluated by Western Blot on normal rat serum.

Western Blot Analysis:
0.05 μg/mL - 1 μg/mL antibody detected fibronectin in 15 μL of normal rat serum.

Target description

262 kDa

Physical form

Antigen Affinity Purified
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Western Blot: Normal rat serum

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rugmani Padmanabhan Iyer et al.
American journal of physiology. Heart and circulatory physiology, 311(3), H822-H836 (2016-08-16)
The purpose of this study was to evaluate the effect of sham surgery in a minimally invasive surgical model of permanent coronary artery occlusion used to generate myocardial infarction (MI) in mice. Adult male C57BL/6J mice (3-6 mo old) were
Ina V Martin et al.
American journal of physiology. Renal physiology, 314(1), F35-F46 (2017-09-15)
Complement factor C5a has two known receptors, C5aR, which mediates proinflammatory effects, and C5L2, a potential C5a decoy receptor. We previously identified C5a/C5aR signaling as a potent profibrotic pathway in the kidney. Here we tested for the first time the
Cláudia Silva Souza et al.
Experimental biology and medicine (Maywood, N.J.), 244(9), 758-769 (2019-05-02)
To date, no studies have been found evaluating the effects of physical exercise on renal function and structure changes in ovariectomized rats with type 1 diabetes. Therefore, this work emerges with an important tool for strengthening and expanding innovative research
Tao Tang et al.
Connective tissue research, 54(1), 5-13 (2012-07-28)
Biglycan (BGN), a small leucine-rich proteoglycan, binds the pro-fibrotic cytokine transforming growth factor β (TGFβ) and inhibits its bioactivity in vitro. Nevertheless, it is controversial whether BGN plays an inhibitory role in vivo. Therefore, the purpose of this study was
Ruiting Wang et al.
Cell biology international, 46(3), 381-390 (2021-12-23)
Mitochondrial homeostasis plays essential role for the proper functioning of the kidney. NADH-ubiquinone oxidoreductase core subunit V1 (NDUFV1) is an enzyme in the complex I of electron transport chain (ETC) in mitochondria. In the present study, we examined the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service