Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500203

Sigma-Aldrich

Anti-Cathepsin K antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CTSK, Anti-CTSO, Anti-CTSO1, Anti-CTSO2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTSK(1513)

General description

Procathepsin K is the inactive proenzyme of cathepsin K. Cathepsin K is a lysosomal cysteine protease and part of the papain cysteine protease family. It is expressed in osteoclasts. The gene encoding this protein is localized on human chromosome 1q21.

Immunogen

Peptide with sequence C-KTHRKQYNNKVDE from the internal region (near N-terminus) of the protein sequence according to NP_000387.1.

Biochem/physiol Actions

Cathepsin K has a role in bone resorption and remodeling. It has a triple helical collagen hydrolase activity. Mutations in the gene encoding this protein have been linked to pycnodysostosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joaquín Bobillo Lobato et al.
Medicina clinica, 145(7), 281-287 (2015-02-11)
Gaucher disease is an inherited disorder caused by deficit of acid β-glucocerebrosidase, responsible for the degradation of glucosylceramide to ceramide and glucose. Although the disorder is primarily hematologic, bone is the second most commonly affected structure. Cathepsin K (CATK) is
Xianglan Huang et al.
Calcified tissue international, 96(5), 373-378 (2015-03-03)
Pycnodysostosis is a rare autosomal recessive skeletal dysplasia characterized by short stature, osteosclerosis, acro-osteolysis, frequent fractures, and skull deformities. Mutation in the gene encoding cathepsin K (CTSK), which is a lysosomal cysteine protease, has been found to be responsible for
Jaime Toral-López et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 59(2), 277-280 (2010-11-26)
Pycnodysostosis, an autosomal recessive skeletal dysplasia, is characterized by short stature, osteosclerosis, delayed cranial suture closure, hypoplastic mandible, acro-osteolysis, hypoplastic clavicle, and dental anomalies. The disorder is caused by CTSK gene defects, a gene localized on 1q21. To describe the
Guangxian Zhao et al.
PloS one, 10(8), e0136093-e0136093 (2015-08-25)
Cysteinyl cathepsin K (CatK) is one of the most potent mammalian collagenases involved in cardiovascular disease. Here, we investigated the clinical predictive value of serum CatK levels in patients with chronic heart failure (CHF). We examined 134 patients with CHF
Adeleke H Aguda et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(49), 17474-17479 (2014-11-26)
Cathepsin K is the major collagenolytic protease in bone that facilitates physiological as well as pathological bone degradation. Despite its key role in bone remodeling and for being a highly sought-after drug target for the treatment of osteoporosis, the mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service