Skip to Content
Merck

B2025

Brilliant Blue G - Colloidal Concentrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Level

storage temp.

room temp

General description

Coomassie Brilliant Blue stain is widely used for the detection of proteins in sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS PAGE). Staining with brilliant blue G is called as blue silver staining due to its improved sensitivity close to silver staining method. Coomassie Brilliant Blue G stain associates with proteins via hydrophobic interaction. It is effective in staining anterior capsule in cornea with relatively less toxic effects.

Application

Brilliant Blue G - Colloidal Concentrate has been used in the staining
  • protein fragments post in vitro pepsin digestion
  • ovarian tumor fluid proteome prior to profiling
  • the transcription factor (TFIID) complex from Saccharomyces cerevisiae

Preparation Note

The suspension will contain 0.1% (w/v) Brilliant Blue G, 0.29 M phosphoric acid and 16% saturated ammonium sulfate after diluting to 1 liter with 800 mL water.

Analysis Note

This product has been tested for suitability on SDS-PAGE.


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Blue silver: a very sensitive colloidal Coomassie G-250 staining for proteome analysis
Candiano G, et al.
Electrophoresis, 25(9), 1327-1333 (2004)
Elena Barbieri et al.
Journal of aging research, 2011, 845379-845379 (2011-06-02)
This study describes mitochondrial behaviour during the C2C12 myoblast differentiation program and proposes a proteomic approach to mitochondria integrated with classical morphofunctional and biochemical analyses. Mitochondrial ultrastructure variations were determined by transmission electron microscopy; mitochondrial mass and membrane potential were
A proteomic signature of ovarian cancer tumor fluid identified by highthroughput and verified by targeted proteomics
Poersch A, et al.
Journal of proteomics, 145(2), 226-236 (2016)



Global Trade Item Number

SKUGTIN
B2025-1EA04061833426524