Skip to Content
Merck
All Photos(3)

Key Documents

S9951

Sigma-Aldrich

Soyasaponin I

Synonym(s):

SCM 3B, Soyasaponin Bb

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H78O18
CAS Number:
Molecular Weight:
943.12
MDL number:
UNSPSC Code:
12352212
NACRES:
NA.25

biological source

Glycine max (soybean)

Assay

≥94% (HPLC)

form

powder

storage condition

protect from light

solubility

ethanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC1(C)C[C@@H](O)[C@@](CC[C@]2(C)C3=CC[C@@]4([H])[C@@]2(C)CC[C@]5([H])[C@]4(C)CC[C@H](O[C@]6([H])O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]6O[C@@]7([H])[C@H](O[C@]8([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]8O)[C@@H](O)[C@@H](O)[C@@H](CO)O7)[C@@]5(CO)C)(C)[C@@]3([H])C

InChI

1S/C48H78O18/c1-21-29(52)31(54)35(58)40(61-21)65-37-32(55)30(53)24(19-49)62-41(37)66-38-34(57)33(56)36(39(59)60)64-42(38)63-28-12-13-45(5)25(46(28,6)20-50)11-14-48(8)26(45)10-9-22-23-17-43(2,3)18-27(51)44(23,4)15-16-47(22,48)7/h9,21,23-38,40-42,49-58H,10-20H2,1-8H3,(H,59,60)/t21-,23-,24+,25+,26+,27+,28-,29-,30-,31+,32-,33-,34-,35+,36-,37+,38+,40-,41-,42+,44+,45-,46+,47+,48+/m0/s1

InChI key

PTDAHAWQAGSZDD-IOVCITQVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Soyasaponin I is an oleanene-type triterpenoid aglycone and is a group B soyasaponin. It is an amphiphilic molecule and is metabolized by intestinal bacteria into sugars and aglycones.

Application

Soyasaponin I has been used:
  • to test its effect on migration and invasion of ovarian cancer cell lines
  • as standard for the quantification of saponins from quinoa and fenugreek samples
  • as L-15 medium supplement for Zebrafish liver cell lines

Biochem/physiol Actions

Antioxidant and anti-inflammatory saponin found in soybean. Attenuates TNBS-induced colitis in mice. The structure consists of a glycoside moiety and triterpene derivative. Sialytransferase inhibitor and renin inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco A Lazo-Vélez et al.
Journal of food science, 81(1), C19-C26 (2015-12-10)
Pseudocereal Chenopodium berlandieri spp. (huauzontle) was evaluated to determine saponin composition. Saponins were evaluated in raw and germinated grains subjected to chemical stress induced by sodium selenite. Analysis by liquid chromatography coupled with ELSD detector revealed the presence of 12
Soyasaponin I and sapongenol B have limited absorption by Caco-2 intestinal cells and limited bioavailability in women
Hu J, et al.
The Journal of Nutrition, 134(8), 1867-1873 (2004)
Cell Recognition Molecule L1 Regulates Cell Surface Glycosylation to Modulate Cell Survival and Migration.
Gang Shi et al.
International journal of medical sciences, 14(12), 1276-1283 (2017-11-07)
San Huang et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 24(3), 232-241 (2017-12-05)
Soaking of legumes results in the loss of macronutrients, micronutrients and phytochemicals. Fibre, protein and phytochemicals found in legumes exert emulsifying activity that may improve the structure and texture of gluten-free bread. The legume soaking water of haricot beans, garbanzo
Pi-Lin Sung et al.
Taiwanese journal of obstetrics & gynecology, 57(2), 255-263 (2018-04-21)
Our previous study has shown that high expression of α2,3-sialytransferase type I was associated with advanced stage serous type epithelial ovarian cancer (EOC). The aim of the current study further attempts to evaluate the altered α 2,3-sialylation on the behavior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service