Skip to Content
Merck
All Photos(3)

Documents

S9132

Sigma-Aldrich

Spiramycin

Synonym(s):

Rovamycin, Formacidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51283208
PubChem Substance ID:
NACRES:
NA.85

form

powder

Quality Level

optical activity

[α]/D -85 to -80° in water (Specific rotation (dry basis))

potency

≥4100 I.U. per mg (dry basis)

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to light yellow

solubility

methanol: soluble

cation traces

heavy metals: ≤20 ppm

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](OC2CCC(C(C)O2)N(C)C)[C@H](C)C[C@H](CC=O)[C@@H]1OC3OC(C)C(OC4CC(C)(O)C(O)C(C)O4)C(C3O)N(C)C

InChI

1S/C43H74N2O14/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3/b13-12+,16-14+/t24-,25-,26?,27?,28?,29+,30?,31-,32+,34?,35?,36?,37?,38?,39+,40+,41?,42?,43?/m1/s1

InChI key

ACTOXUHEUCPTEW-JMRHEKERSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Spiramycin is a macrolide antibiotic that is commonly used to treat infections of soft tissues. It has been used to treat bronchopulmonary infections in adults and has been used to study septicemia in mice .

Biochem/physiol Actions

Spiramycin is a 16-membered ring macrolide antibiotic from Streptomyces ambofaciens. It inhibits bacterial protein synthesis at the level of peptidy-tRNA dissociation from ribosomes. It is mainly used against Gram-positive bacteria.

Quality

A mixture of Spiramycin I, II and III.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Rolin et al.
Pathologie-biologie, 34(5), 476-478 (1986-05-01)
Experimental septicemia was induced in mice by intraperitoneal injection of 10 to 100 lethal doses of Staphylococcus aureus and Streptococcus pneumoniae. Animals were treated by a mixture of adipic acid and spiramycin (subcutaneous route) or by spiramycin base (oral route)
Acute bronchopulmonary infections: treatment with i.v. spiramycin.
F Vachon and S Kernbaum
Chemotherapia, 6, 282-285 (1987)
Yonca Acikgoz et al.
Pediatric transplantation, 16(4), E115-E119 (2011-02-16)
Cryptosporidium is an intracellular protozoan parasite that causes gastroenteritis in human. In immunocompromised individuals, cryptosporidium causes far more serious disease. There is no effective specific therapy for cryptosporidiosis, and spontaneous recovery is the rule in healthy individuals. However, immunocompromised patients
Ming-juan Wang et al.
Journal of chromatography. A, 1217(42), 6531-6544 (2010-09-17)
Investigation of acetylspiramycin (ASPM) and its related substances was carried out using a reversed-phase liquid chromatography/tandem mass spectrometry method. The identification of impurities in the ASPM complex was performed with a quadrupole ion trap mass spectrometer, with an electrospray ionization
Severe congenital toxoplasmosis due to a Toxoplasma gondii strain with an atypical genotype: case report and review.
Laurence Delhaes et al.
Prenatal diagnosis, 30(9), 902-905 (2010-06-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service