Skip to Content
Merck
All Photos(1)

Key Documents

32071

Supelco

Nafcillin sodium salt

VETRANAL®, analytical standard

Synonym(s):

(2S,5R,6R)-6-{[(2-Ethoxy-1-naphthalenyl)carbonyl]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium salt, 6-(2-Ethoxy-1-naphthamido)penicillin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N2NaO5S
CAS Number:
Molecular Weight:
436.46
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[Na+].S1[C@H]2N([C@H](C1(C)C)C(=O)O)C(=O)[C@H]2\N=C(/[O-])\c3c4c(ccc3OCC)cccc4

InChI

1S/C21H22N2O5S.Na/c1-4-28-13-10-9-11-7-5-6-8-12(11)14(13)17(24)22-15-18(25)23-16(20(26)27)21(2,3)29-19(15)23;/h5-10,15-16,19H,4H2,1-3H3,(H,22,24)(H,26,27);/q;+1/p-1/t15-,16+,19-;/m1./s1

InChI key

AYAPZOUDXCDGIF-FRFVDRIFSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam
Nafcillin is a penicillin antibiotic used mostly in treating mastitis of cows.

Application

Nafcillin sodium salt (NF-PC) has been used as working standard in HPLC analysis to determine β-lactam antibiotics from milk samples and in LC–MS–MS analysis to determine NF-PC in bovine tissues.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marin L Schweizer et al.
BMC infectious diseases, 11, 279-279 (2011-10-21)
The high prevalence of methicillin-resistant S. aureus (MRSA) has led clinicians to select antibiotics that have coverage against MRSA, usually vancomycin, for empiric therapy for suspected staphylococcal infections. Clinicians often continue vancomycin started empirically even when methicillin-susceptible S. aureus (MSSA)
Mohan Pammi et al.
Pediatric research, 70(6), 578-583 (2011-08-23)
Biofilm infections are frequently caused by Staphylococcus epidermidis, are resistant to antimicrobial agents, and adversely affect patient outcomes. We evaluated farnesol (FSL), the Candida quorum-sensing molecule, on S. epidermidis biofilms, in vitro and in vivo. We evaluated ED50, ED75, and
Pierre Tattevin et al.
Research in microbiology, 160(3), 187-192 (2009-04-16)
Growth of fluoroquinolone (FQ)-susceptible methicillin-resistant Staphylococcus aureus (MRSA) in the presence of sub-MIC FQ has been shown to enhance methicillin resistance in traditional nosocomial MRSA isolates. We aimed to confirm this phenomenon in nine community-associated MRSA (CA-MRSA) clinical isolates, and
Steven N Leonard
PloS one, 7(7), e42103-e42103 (2012-08-01)
Continued pressure from glycopeptide use has led to non-susceptible strains of Staphylococcus aureus including heterogeneously vancomycin-intermediate S. aureus (hVISA). Infections with hVISA are associated with poor patient outcomes, thus incentivizing novel treatments. Evidence suggests that vancomycin and anti-staphylococcal penicillin susceptibility
Craig B Whitman et al.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 69(12), 1049-1053 (2012-05-31)
To report a case of acute interstitial nephritis (AIN) secondary to nafcillin. A 55-year-old Hispanic man (height, 63 in.; weight, 61.2 kg) with a history of deep vein thrombosis in the right lower extremity, hypertension, hyperlipidemia, and hepatitis C infection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service