Skip to Content
Merck
All Photos(1)

Key Documents

S6272

Sigma-Aldrich

Substance P Fragment 1-7

≥97% (HPLC)

Synonym(s):

Arg-Pro-Lys-Pro-Gln-Gln-Phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H65N13O10
CAS Number:
Molecular Weight:
900.04
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Substance P Fragment 1-7, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥65%

color

white

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

SMILES string

NCCCCC(NC(=O)C1CCCN1C(=O)C(N)CCCNC(N)=N)C(=O)N2CCCC2C(=O)NC(CCC(N)=O)C(=O)NC(CCC(N)=O)C(=O)NC(Cc3ccccc3)C(O)=O

InChI

1S/C41H65N13O10/c42-19-5-4-12-28(51-37(60)30-13-7-21-53(30)38(61)25(43)11-6-20-48-41(46)47)39(62)54-22-8-14-31(54)36(59)50-27(16-18-33(45)56)34(57)49-26(15-17-32(44)55)35(58)52-29(40(63)64)23-24-9-2-1-3-10-24/h1-3,9-10,25-31H,4-8,11-23,42-43H2,(H2,44,55)(H2,45,56)(H,49,57)(H,50,59)(H,51,60)(H,52,58)(H,63,64)(H4,46,47,48)

InChI key

KPHDBQWTCKBKIL-UHFFFAOYSA-N

Gene Information

human ... TAC1(6863)

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Phe

Biochem/physiol Actions

Fragment that gives depressor and bradycardic effects when applied to the nucleus tractus solitarius

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milad Botros et al.
Peptides, 27(4), 753-759 (2005-10-12)
Endomorphin-1 (EM-1) and endomorphin-2 (EM-2) represent two opioid active tetrapeptides with high affinity and selectivity for the mu-opioid (MOP) receptor. Both EM-1 and EM-2 exhibit strong inhibition of pain signals in the central nervous system (CNS). In contrast to these
Qin Zhou et al.
Peptides, 25(11), 1951-1957 (2004-10-27)
The aminoterminal fragment of substance P (SP), SP(1-7), has been suggested to modulate the expression of opiate tolerance and withdrawal behaviors in rodents. However, the mechanism of this effect is not yet clarified. Using a rat model we have previously
Qin Zhou et al.
Neuroscience letters, 320(3), 117-120 (2002-02-20)
The biologically active substance P (SP) N-terminal metabolite SP(1-7) has been reported to modulate several neural processes such as learning, locomotor activity and reaction to opioid withdrawal. Although all these processes are believed to be associated with dopaminergic transmission no
Qin Zhou et al.
Peptides, 32(8), 1661-1665 (2011-07-19)
The N-terminal substance P fragment SP(1-7) is known to modulate hyperalgesia and opioid withdrawal in animal models. This study examined the effects of intraperitoneal (i.p.) injections of SP(1-7) on chronic morphine tolerance and on the levels of dynorphin B (DYN
Anna Carlsson et al.
European journal of pharmacology, 626(2-3), 250-255 (2009-10-20)
We have recently explored the role of the tachykinin substance P neuroactive fragment substance P(1-7) in the mediation of anti-inflammatory effects using a blister model in the rat paw (Wiktelius et al., 2006). We observed that this heptapeptide induced a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service