Skip to Content
Merck
All Photos(1)

Key Documents

S4195

Sigma-Aldrich

β-Secretase human

recombinant, expressed in HEK 293 cells (C-terminal FLAG tagged), extracellular domain, ≥10,000 units/mg protein, ≥90% (SDS-PAGE), buffered aqueous solution

Synonym(s):

Asp-2, BACE1, Memapsin-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in HEK 293 cells (C-terminal FLAG tagged)

Quality Level

Assay

≥90% (SDS-PAGE)

form

buffered aqueous solution

specific activity

≥10,000 units/mg protein

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... BACE1(23621)

Looking for similar products? Visit Product Comparison Guide

Application

Human β-secretase or BACE1 has been used to study the binding reaction between amyloid precursor protein and β-secretase. It has also been used in BACE1 assay.

Biochem/physiol Actions

BACE1 (β−Secretase or β−Site APP-Cleaving Enzyme) is a transmembrane protease responsible for the β site cleavage of the amyloid precursor protein (APP) to produce amyloid β peptide (Aβ). The accumulation of Aβ in the brain is a primary cause for the progression of Alzheimer′s, therefore BACE1 is a target for inhibitor drug discovery.
BACE1 (β-site APP-cleaving enzyme 1) is a transmembrane aspartyl protease responsible for cleaving the amyloid precursor protein (APP). BACE1-mediated cleavage of amyloid precursor protein (APP) is the first step during the generation of pathogenic amyloid-β peptides, hence making it the major drug target for Alzheimer′s disease (AD). Increased levels of enzymatic activity of BACE1 was found in cerebrospinal fluid from patients having both mild cognitive impairment (MCI) and AD, thereby suggesting the importance of BACE1 enzymatic activity in conversion of MCI to AD.
Transmembrane protease responsible for the β site cleavage of the amyloid precursor protein (APP) to produce amyloid β peptide.

Unit Definition

One unit will hydrolyze 1.0 picomole of 7-methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid β/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)-Lys-Arg-Arg amide substrate per 1 minute at pH 4.5 at 37 °C.

Physical form

Solution in 20 mM Hepes, pH 7.4, 125 mM NaCl.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platelet-Derived Secreted Amyloid-Precursor Protein-? as a Marker for Diagnosing Alzheimer?s Disease
Current neurovascular research (2013)
Roland Nicsanu et al.
Journal of Alzheimer's disease : JAD, 87(1), 433-441 (2022-03-12)
Beta-site APP cleaving enzyme 1 (BACE1) is the rate-limiting enzyme in amyloid-β (Aβ) plaques formation. BACE1 activity is increased in brains of patients with Alzheimer's disease (AD) and mild cognitive impairment (MCI) and plasma levels of BACE1 appears to reflect
Function, therapeutic potential and cell biology of BACE proteases: current status and future prospects.
Vassar R
Journal of Neurochemistry (2014)
Carlo Cervellati et al.
GeroScience, 42(1), 159-167 (2019-11-21)
Beta-secretase (BACE1) is a key enzyme in the formation of amyloid-β; its activity/concentration is increased in brain and cerebrospinal fluid of patients with late-onset Alzheimer's disease (LOAD). Since BACE1 was found also in blood, we evaluated its potential as peripheral
Platelet-Derived Secreted Amyloid-Precursor Protein-? as a Marker for Diagnosing Alzheimer?s Disease
Josef M and Christian H
Current neurovascular research (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service