Skip to Content
MilliporeSigma

Skip To

CS0010

β-Secretase (BACE1) Activity Detection Kit (Fluorescent)

1 kit sufficient for 250 reactions

Synonym(s):

BACE1 Activity Detection Kit (Fluorescent)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
$1,900.00

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503

$1,900.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

usage

 kit sufficient for 250 reactions

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BACE1(23621)

Application

The kit provides all the reagents required for an efficient detection of BACE1 activity. It contains an enzyme to be used for screening for potential BACE1 inhibitors. The assay is based on the fluorescence resonance energy transfer (FRET) method in which the fluorescence signal enhancement is observed after substrate cleavage by BACE1.

Biochem/physiol Actions

BACE1 is a transmembrane protease responsible for the β site cleavage of the amyloid precursor protein (APP) to produce amyloid β peptide (Aβ). The accumulation of Aβ in the brain is a primary cause for the progression of Alzheimer′s. BACE1 is a target for inhibitor drug discovery.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S4195RAB0623AB5832
Gene Information

human ... BACE1(23621)

Gene Information

human ... BACE1(23621)

Gene Information

human ... BACE1(23621)

Gene Information

human ... BACE1(23621)

Quality Level

300

Quality Level

200

Quality Level

-

Quality Level

100

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

-

usage

 kit sufficient for 250 reactions

usage

-

usage

-

usage

-


Kit Components Only

Product No.
Description

  • Fluorescent Assay Buffer 50 mL

  • Stop Solution 15 mL

  • Substrate (MOCA-SEV-NL-DAEFR-DNP-RR) 500 μL

  • Assay Standard 140 μL

  • BACE1 (β−Secretase) 300 units 100 μL

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. what is the concentration of BACE1 enzyme in micromolar?

    1 answer
    1. The B9059 BACE-1 product included in the kit, serves as a positive control. The Technical Bulletin notes that the concentration of this item is approximate 3 units/microliter. This material has not been tested for purity. Therefore, a molar concentration cannot be provided.

      Helpful?

Reviews

No rating value

Active Filters