Skip to Content
Merck
All Photos(2)

Key Documents

C9584

Sigma-Aldrich

Carboxypeptidase B from porcine pancreas

lyophilized powder

Synonym(s):

Peptidyl-L-lysine(L-arginine) hydrolase, Protaminase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54

biological source

Porcine pancreas

grade

Proteomics Grade

form

lyophilized powder

specific activity

≥125 units/mg protein

mol wt

34,000 Da± 600

purified by

affinity chromatography

composition

Protein, 40-70%

foreign activity

carboxypeptidase A ≤1%
chymotrypsin ≤0.1%
trypsin ≤5%

storage temp.

−20°C

SMILES string

S(Cc3ccccc3)C(=O)C(NC(=O)C2N(CCC2)C(=O)C(NC(=O)C(NC(=O)CCC(=O)O)C)C)Cc1ccccc1

InChI

1S/C31H38N4O7S/c1-20(32-26(36)15-16-27(37)38)28(39)33-21(2)30(41)35-17-9-14-25(35)29(40)34-24(18-22-10-5-3-6-11-22)31(42)43-19-23-12-7-4-8-13-23/h3-8,10-13,20-21,24-25H,9,14-19H2,1-2H3,(H,32,36)(H,33,39)(H,34,40)(H,37,38)

InChI key

TWURVFFNODFJBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carboxypeptidase B has been used in a study to develop a non-invasive pregnancy assay for use in both captive and wild polar bears. Carboxypeptidase B has been used in a study that identified new potential biomarkers of acute pancreatitis.
The enzyme from Sigma has been used to develop homogeneous time-resolved fluorescence (HTRF) assay for measuring carboxypeptidase B activity in a miniaturized high-throughput screening format. It has been used to evaluate the impact of the C-terminal lysine(s) in human plasminogen binding to Bifidobacterium. The effect of treatment with carboxypeptidase B, which is a C-terminal lysine-specific endopeptidase, is measured using flow cytometry analysis.

Biochem/physiol Actions

Carboxypeptidase B is a proteolytic enzyme capable of rapidly hydrolyzing peptide bonds to release certain carboxyl-terminal basic amino acids from peptides and proteins. Its molecular mass is 34,300±600 Da. It contains one non-dialyzable gram atom of zinc per mole. The enzyme activity is inhibited by metal chelating agents 1, 10-phenanthroline, 8-hydroxyquinoline-5-sulfonic acid, and 2,2′-dipyridyl.

Packaging

Package size based on protein content

Unit Definition

One unit will hydrolyze 1.0 μmole of hippuryl-L-arginine per min at pH 7.65 at 25 °C.

Physical form

Contains HEPES buffer salts, zinc chloride and carbohydrate

Preparation Note

Treated with protease inhibitor, AEBSF, to eliminate serine protease activity.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Ferrer et al.
Analytical biochemistry, 317(1), 94-98 (2003-05-06)
An epitope-unmasking, homogeneous time-resolved fluorescence (HTRF) assay has been developed for measuring carboxypeptidase B (CPB) activity in a miniaturized high-throughput screening format. The enzyme substrate (biotin-RYRGLMVGGVVR-OH) is cleaved by CPB at the C terminus, causing release of the C-terminal Arg
Laboratory diagnosis of acute pancreatitis: in search of the Holy Grail.
Lippi, G. et al.
Clinical Laboratory Science : Journal of the American Society For Medical Technology, 49, 18-31 (2012)
Marco Candela et al.
Journal of bacteriology, 189(16), 5929-5936 (2007-06-15)
Bifidobacteria constitute up to 3% of the total microbiota and represent one of the most important health-promoting bacterial groups of the human intestinal microflora. The presence of Bifidobacterium in the human gastrointestinal tract has been directly related to several health-promoting
Andrew Lee et al.
Langmuir : the ACS journal of surfaces and colloids, 27(18), 11560-11574 (2011-08-13)
This article describes the use of capillary electrophoresis (CE) to examine the influence of different cations (C(+); C(+) = Na(+) and tetra-n-alkylammonium, NR(4)(+), where R = Me, Et, Pr, and Bu) on the rates of denaturation of bovine carbonic anhydrase
E Curry et al.
Theriogenology, 78(2), 308-314 (2012-04-28)
Currently, there is no method of accurately and non-invasively diagnosing pregnancy in polar bears. Specific proteins may exhibit altered profiles in the feces of pregnant bears, but predicting appropriate candidate proteins to investigate is speculative at best. The objective of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service