Skip to Content
Merck
All Photos(1)

Key Documents

PZ0185

Sigma-Aldrich

PF-431396 hydrate

≥98% (HPLC)

Synonym(s):

N-[2-[[[2-[(2,3-dihydro-2-oxo-1H-indol-5-yl)amino]-5-(trifluoromethyl)-4-pyrimidinyl]amino]methyl]phenyl]-N-methyl-Methanesulfonamide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21F3N6O3S · xH2O
CAS Number:
Molecular Weight:
506.50 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to blue-gray

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O.CN(c1ccccc1CNc2nc(Nc3ccc4NC(=O)Cc4c3)ncc2C(F)(F)F)S(C)(=O)=O

InChI

1S/C22H21F3N6O3S.H2O/c1-31(35(2,33)34)18-6-4-3-5-13(18)11-26-20-16(22(23,24)25)12-27-21(30-20)28-15-7-8-17-14(9-15)10-19(32)29-17;/h3-9,12H,10-11H2,1-2H3,(H,29,32)(H2,26,27,28,30);1H2

InChI key

XSXCARUUZVMRGX-UHFFFAOYSA-N

Application

PF-431396 hydrate has been used as a dual inhibitor of proline-rich tyrosine kinase 2 (PYK2) / focal adhesion kinase (FAK) inhibitor to study its effects on phosphorylation of Yes-associated protein (YAP) at Ser127 and steady state of transcriptional coactivator with PDZ-binding motif (TAZ) . It has also been used as a PYK2/FAK inhibitor to study its effects on protein kinase A activation in human sperm samples .

Biochem/physiol Actions

PF-431396 is a potent inhibitor of PYK2 and FAK kinases (IC50 = 11 and 1.5 nM, respectively). PF-431396 increases bone formation and protects against bone loss in ovariectomized rats.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Liu et al.
Neuroreport, 31(10), 754-761 (2020-05-27)
The aim of this study was to investigate the effects of histone deacetylase-6 (HDAC6) on the functional and pathological changes of the amyloid beta (Aβ)-induced cognitive dysfunction rats by regulating protein tyrosine kinase 2 beta (PTK2B). Ninety Sprague Dawley rats
Seyedeh Fatemeh Razavipour et al.
Nature communications, 15(1), 5152-5152 (2024-06-18)
In many cancers, a stem-like cell subpopulation mediates tumor initiation, dissemination and drug resistance. Here, we report that cancer stem cell (CSC) abundance is transcriptionally regulated by C-terminally phosphorylated p27 (p27pT157pT198). Mechanistically, this arises through p27 co-recruitment with STAT3/CBP to
Jialie Luo et al.
Immunity, 56(7), 1533-1547 (2023-06-25)
The crosstalk between the immune and neuroendocrine systems is critical for intestinal homeostasis and gut-brain communications. However, it remains unclear how immune cells participate in gut sensation of hormones and neurotransmitters release in response to environmental cues, such as self-lipids
PYK2 negatively regulates the Hippo pathway in TNBC by stabilizing TAZ protein
Kedan A, et al.
Cell Death & Disease, 9(10), 1-15 (2018)
Evidence for the involvement of proline-rich tyrosine kinase 2 in tyrosine phosphorylation downstream of protein kinase A activation during human sperm capacitation
Battistone M A, et al.
Molecular Human Reproduction, 20(11), 1054-1066 (2014)

Articles

The focal adhesion kinase (FAK) is a cytoplasmic protein tyrosine kinase. FAK has been implicated as a downstream signaling molecule that functions in the control of several integrin-regulated biological processes.

The focal adhesion kinase (FAK) is a cytoplasmic protein tyrosine kinase. FAK has been implicated as a downstream signaling molecule that functions in the control of several integrin-regulated biological processes.

The focal adhesion kinase (FAK) is a cytoplasmic protein tyrosine kinase. FAK has been implicated as a downstream signaling molecule that functions in the control of several integrin-regulated biological processes.

The focal adhesion kinase (FAK) is a cytoplasmic protein tyrosine kinase. FAK has been implicated as a downstream signaling molecule that functions in the control of several integrin-regulated biological processes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service