Skip to Content
Merck
All Photos(1)

Key Documents

P6486

Millipore

EZview Red Protein A Affinity Gel

Synonym(s):

EZview Affinity Gel, Protein A Affinity Gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56
Pricing and availability is not currently available.

form

liquid

Quality Level

analyte chemical class(es)

proteins (Immunoglobulins of various mammalian species)

technique(s)

affinity chromatography: suitable
immunoprecipitation (IP): suitable

capacity

~4 mg binding capacity (rabbit IgG)

shipped in

wet ice

storage temp.

−20°C

General description

EZview Red Protein A Affinity Gel is a highly visible, red colored Protein A affinity gel. The affinity resin contains Staphylococcus aureus Protein A covalently attached to crosslinked 4% agarose beads. The affinity gel is used to bind the FC portion of the IgG of an antibody to samples, such as cell lysates and tissue. The immune complexes, bound to the EZview Red Protein A Affinity Gel, are recovered by centrifugation. Red dye enhances visibility for more efficient results.

Application

EZview Red Protein A Affinity Gel has been used for purification of proteins in the preparation of immunoprecipitation[1][2] and chromatin immunoprecipitation (ChIP)[3] assays.
When performing small-scale affinity capture, such as immunoprecipitation, the affinity matrix is difficult to see in the microcentrifuge tubes. Accidental aspiration of the resin leads to quantitative variability in results. The EZview Red Affinity Gels greatly reduce the risk of pellet loss. EZview resins perform as well as conventional non-colored affinity gels, but allow the user to easily differentiate pellet from supernatant. This correlates to more accurate data because less protein is lost.

Features and Benefits

  • Increased visibility - Red color reduces risk of incidental aspiration
  • Improved recovery of target protein by reduced accidental loss
  • Higher reproducibility - More consistent yields

Physical form

1:1 (v/v) suspension in PBS containing 50% glycerol and 15 ppm Kathon
50% slurry in PBS with 50%Glycerol and 0.0015%

Storage and Stability

EZview Red Protein A Affinity Gel should be stored in 50% glycerol at -20 °C. This product is stable for at least one year when stored as indicated. Do not freeze in the absence of glycerol.

Legal Information

EZview is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Matsunaga et al.
Microbiology (Reading, England), 152(Pt 12), 3777-3786 (2006-12-13)
Leptospirosis is a widespread zoonosis caused by invasive spirochaetes belonging to the genus Leptospira. Pathogenic leptospires disseminate via the bloodstream to colonize the renal tubules of reservoir hosts. Little is known about leptospiral outer-membrane proteins expressed during the dissemination stage
NF-?B directly regulates ?-arrestin-1 expression and forms a negative feedback circuit in TNF-a?induced cell death
Li J, et al.
Faseb Journal (2018)
A laboratory practical illustrating the use of the ChIP-qPCR method in a robust model: Estrogen receptor alpha immunoprecipitation using Mcf-7 culture cells
Lacazette E
BioChemistry: An Indian Journal (2017)
Hsp90 Co-chaperone p23 contributes to dopaminergic mitochondrial stress via stabilization of PHD2: Implications for Parkinson's disease.
Rane A, et al.
Neurotoxicology (2018)
Katriina Kahlos et al.
Molecular and cellular biochemistry, 254(1-2), 47-54 (2003-12-17)
We previously reported that exposure to exogenous nitric oxide (NO) causes diminished expression of thioredoxin/thioredoxin reductase, a critical component of the redox system that regulates the functions of redox-sensitive enzymes, receptors, and transcription factors. Here we examined the role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service