Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.56
UNSPSC Code:
41106500
form
suspension
Quality Level
shelf life
1 yr
technique(s)
western blot: suitable
matrix
4% agarose
pH
7.2
capacity
~10 μg(biotin per ml of packed gel)
shipped in
wet ice
storage temp.
2-8°C
General description
The EZ View Red Streptavidin Affinity gel is composed of streptavidin, covalently attached to
cyanogen bromide-activated 4% agarose beads. It is designed to capture (pull-down) the biotinylated target molecules, such as proteins, peptides, antibodies, nucleic acids, lectins, receptors and ligands.
cyanogen bromide-activated 4% agarose beads. It is designed to capture (pull-down) the biotinylated target molecules, such as proteins, peptides, antibodies, nucleic acids, lectins, receptors and ligands.
Application
Sutitable for use with immunoprecipitation, western blotting,and enzyme assays.
When performing small-scale affinity capture, such as immunoprecipitation, the affinity matrix is difficult to see in the microcentrifuge tubes. Accidental aspiration of the resin leads to quantitative variability in results. The EZview™ Red Affinity Gels greatly reduce the risk of pellet loss. EZview™ resins perform as well as conventional non-colored affinity gels, but allow the user to easily differentiate pellet from supernatant. This correlates to more accurate data because less protein is lost.
Features and Benefits
- Increased visibility - Red color reduces risk of incidental aspiration
- Improved recovery of target protein by reduced accidental loss
- Higher reproducibility - More consistent yields
Physical form
1:1 (v/v) suspension in PBS containing 50% glycerol and 15 ppm Kathon
Legal Information
EZview is a trademark of Sigma-Aldrich Co. LLC
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Alicia M Bostwick et al.
Biochemical and biophysical research communications, 528(3), 580-585 (2020-06-09)
Mammalian cells contain genetic information in two compartments, the nucleus and the mitochondria. Mitochondrial gene expression must be coordinated with nuclear gene expression to respond to cellular energetic needs. To gain insight into the coordination between the nucleus and mitochondria
Suet Yin Sarah Fung et al.
Current biology : CB, 27(1), 78-86 (2016-12-13)
After cleavage furrow ingression during cytokinesis, nascent daughter cells remain connected by an intercellular bridge (ICB) and the midbody [1, 2]. The midbody becomes an assembly platform for ESCRT complexes that split apart the plasma membrane (PM) anchored to the
Katarzyna M Zientara-Rytter et al.
Cells, 11(1) (2022-01-12)
Pex11, an abundant peroxisomal membrane protein (PMP), is required for division of peroxisomes and is robustly imported to peroxisomal membranes. We present a comprehensive analysis of how the Pichia pastoris Pex11 is recognized and chaperoned by Pex19, targeted to peroxisome
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E5529-1ML | 04061822788947 |
| E5529-5X1ML | 04061822788954 |