Skip to Content
Merck
All Photos(5)

Key Documents

P4749

Sigma-Aldrich

Anti-PADI4 (N-terminal) antibody produced in rabbit

enhanced validation

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PAD4, Anti-PADI5, Anti-PDI4, Anti-Peptidyl Arginine Deaminase Type IV, Anti-Peptidyl Arginine Deaminase V

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoprecipitation (IP): 2-4 μL using DMSO-treated HL60 cells
microarray: suitable
western blot (chemiluminescent): 1:500-1:1,000 using DMSO-treated HL60 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PADI4(23569)

General description

PAD4 (also designated PADI4 or PAD type IV), one of the members of the PAD family. PAD4 was initially named PAD5. However, since it is closely related to rodent PAD4 it was renamed human PAD4. The most noticeable difference between the isotypes is their tissue-specific expression. Expression of the PAD4 gene is tightly linked to myeloid differentiation. It is expressed in granulocytes and monocytes and expressed in the nucleus.
Peptidyl arginine deiminase 4 (PADI4) is one of the four PADIs found in humans. The protein consists of 663 amino acids. PADI4 gene is localized to human chromosome 1p36. It is expressed in hematopoietic tissues, such as spleen, thymus, peripheral blood leucocytes, fetal liver and bone marrow.

Immunogen

synthetic peptide corresponding to amino acids 1-15 of human PADI4, conjugated to KLH via a C-terminal added cysteine residue. The immunizing peptide is specific to human.

Application

Anti-PADI4 (N-terminal) antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.
Anti-PADI4 (N-terminal) antibody produced in rabbit has been used in immunohistochemistry and immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Biochem/physiol Actions

PADs (also known as PADIs), peptidylarginine deiminases, are a family of enzymes, which convert protein-bound arginine residues to citrulline residues in the presence of calcium ions, a process termed deimination. Among the 5 isoforms of PADs, only PAD4 has been identified as an RA-susceptibility gene.
Peptidyl arginine deiminase 4 (PADI4) is responsible for the post-translational conversion of peptidylarginine to citrulline, in the presence of calcium ions. In individuals with rheumatoid arthritis (RA), this gene is expressed in hematological cells and synovial tissues, and variant in this gene is linked with susceptibility to RA. The expression of this protein is linked with DNA hypermethylation in acute promyelocytic leukemia (APL).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptidylarginine deiminase 1-catalyzed histone citrullination is essential for early embryo development
Xiaoqian Z
Scientific Reports (2016)
Sunish Mohanan et al.
Frontiers in immunology, 4, 67-67 (2013-03-20)
PAD4-mediated hypercitrullination of histone H4 arginine 3 (H4R3) has been previously found to promote the formation of Neutrophil Extracellular Traps in inflamed tissues and the resulting histone H4 citrulline 3 (H4Cit3) modification is thought to play a key role in
Protein arginine deiminase 4 (PAD4): current understanding and future therapeutic potential
Jones J, et al.
Current Opinion in Drug Discovery & Development, 12(5), 616-616 (2009)
Potential Role for PAD2 in Gene Regulation in Breast Cancer Cells
Brian D
PLoS ONE (2012)
Functional haplotypes of PADI4, encoding citrullinating enzyme peptidylarginine deiminase 4, are associated with rheumatoid arthritis.
Suzuki A
Nature Genetics (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service