Skip to Content
Merck
All Photos(4)

Documents

O7753

Sigma-Aldrich

Oxaloacetic acid

powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)

Synonym(s):

2-Oxosuccinic acid, Ketosuccinic acid, Oxalacetic acid, Oxobutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2COCOOH
CAS Number:
Molecular Weight:
132.07
Beilstein:
1705475
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

product line

BioReagent

Assay

≥97% (HPLC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

solubility

H2O: 100 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)CC(=O)C(O)=O

InChI

1S/C4H4O5/c5-2(4(8)9)1-3(6)7/h1H2,(H,6,7)(H,8,9)

InChI key

KHPXUQMNIQBQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxaloacetic acid is a dicarboxylic acid that is present in every cell in the body. It is a metabolic intermediate of the tricarboxylic acid (TCA) cycle and gluconeogenesis.

Application

Oxaloacetic acid has been used to evaluate its effect on neuromuscular function and lifespan in amyotrophic lateral sclerosis (ALS) model superoxide dismutase 1 (SOD1) G93A mice. It has also been used as a supplement in Dulbecco′s modified Eagle′s medium (DMEM) to culture chicken fibroblast (DF-1) cells and chemically immortalized leghorn male hepatoma (LMH) cells for functional validation experiments.

Biochem/physiol Actions

Oxaloacetic acid plays a vital role in central metabolism. It inhibits succinate dehydrogenase and is a key regulator of mitochondrial metabolism.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian D Fink et al.
The Journal of biological chemistry, 293(51), 19932-19941 (2018-11-06)
We recently reported a previously unrecognized mitochondrial respiratory phenomenon. When [ADP] was held constant ("clamped") at sequentially increasing concentrations in succinate-energized muscle mitochondria in the absence of rotenone (commonly used to block complex I), we observed a biphasic, increasing then
E Esenmo et al.
The American journal of physiology, 263(1 Pt 1), E36-E41 (1992-07-01)
Estimating the rate of hepatic gluconeogenesis in vivo from the incorporation of 14C from 14CO2 into glucose requires determination of the rates in liver of equilibration of oxaloacetate with fumarate, conversion of oxaloacetate to phosphoenolpyruvate (PEP), and conversion of PEP
Eva Brekke et al.
Glia, 60(1), 147-158 (2011-11-05)
Pyruvate carboxylation (PC) is thought to be the major anaplerotic reaction for the tricarboxylic acid cycle and is necessary for de novo synthesis of amino acid neurotransmitters. In the brain, the main enzyme involved is pyruvate carboxylase, which is predominantly
Anthony Gandin et al.
Plant & cell physiology, 53(9), 1627-1637 (2012-08-01)
In higher plants, the mitochondrial electron transport chain has non-phosphorylating alternative pathways that include the alternative terminal oxidase (AOX). This alternative pathway has been suggested to act as a sink for dissipating excess reducing power, minimizing oxidative stress and possibly
Zhitao Wang et al.
Viruses, 13(6) (2021-07-03)
Gallid alpha-herpesvirus 1, also known as avian infectious laryngotracheitis virus (ILTV), continues to cause huge economic losses to the poultry industry worldwide. Similar to that of other herpesvirus-encoded proteins, the expression of viral genes encoded by ILTV is regulated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service