Skip to Content
Merck
All Photos(3)

Documents

75892

Sigma-Aldrich

α-Ketoglutaric acid disodium salt dihydrate

≥98.0% (dried material, NT)

Synonym(s):

2-Oxoglutaric acid disodium salt, Sodium α-ketoglutarate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2COCOONa · 2H2O
CAS Number:
Molecular Weight:
226.09
Beilstein:
5658566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (dried material, NT)

form

powder

impurities

~16% water

SMILES string

O.O.[Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O

InChI

1S/C5H6O5.2Na.2H2O/c6-3(5(9)10)1-2-4(7)8;;;;/h1-2H2,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2

InChI key

HSSWOFWCOAQLHZ-UHFFFAOYSA-L

General description

α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) is the hydrated disodium salt of 2-oxoglutaric acid.

Application

α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) may be used as starting reagent for the synthesis of deuteriated 2-oxo[3,3-2H2]glutarate disodium salt.
α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kisakurek MV.
Hominatio: An International Tribute to Albert Eschenmoser, 2557-2557 (2001)
Piotr Dobrowolski et al.
Nutrition (Burbank, Los Angeles County, Calif.), 29(3), 556-561 (2012-12-12)
Proton-pump inhibitors, such as omeprazole, are widely used in the prevention and treatment of gastroesophageal diseases. However, an association between proton-pump inhibitors and the increased risk of bone fractures has been observed, especially in patients treated for extended periods. Conversely
Li-Li Chen et al.
Journal of molecular modeling, 19(4), 1919-1927 (2013-01-23)
Bacillus subtilis natto is widely used in industry to produce natto, a traditional and popular Japanese soybean food. However, during its secondary fermentation, high amounts of ammonia are released to give a negative influence on the flavor of natto. Glutamate
R V Durán et al.
Oncogene, 32(38), 4549-4556 (2012-10-23)
Hypoxia-inducible factor (HIF) prolyl hydroxylases (PHDs) are α-ketoglutarate (αKG)-dependent dioxygenases that function as cellular oxygen sensors. However, PHD activity also depends on factors other than oxygen, especially αKG, a key metabolic compound closely linked to amino-acid metabolism. We examined the
Sylwia Magiera et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 919-920, 20-29 (2013-02-16)
A simple, accurate and sensitive liquid chromatography tandem mass spectrometry (LC-MS/MS) method was developed and validated for the quantitation of α-ketoglutaric acid (α-KG), L-carnitine (L-CAR) and acetyl-L-carnitine (acetyl-L-CAR) in human urine as potential biomarkers of cardiovascular disease. The separation was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service