Skip to Content
Merck
All Photos(4)

Documents

P8574

Sigma-Aldrich

Sodium pyruvate

BioXtra, ≥99%

Synonym(s):

α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
Beilstein:
3568341
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

product line

BioXtra

Quality Level

Assay

≥99%

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

mp

>300 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

DAEPDZWVDSPTHF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium pyruvate has been used as a component:
  • of cutting solution for rat brain tissue slice preparation
  • of M-199 medium for the maturation of oocytes
  • of Krebs-Henseleit buffer for plasma incubation

Biochem/physiol Actions

Sodium pyruvate based therapeutic regimen is effective for treating mitochondrial diseases. It decreases tissue damage associated with the pathophysiology of brain damage, haemorrhagic shock and ischemia-reperfusion injury models.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imaging calcium concentration dynamics in small neuronal compartments
Yasuda R, et al.
Science's STKE : Signal Transduction Knowledge Environment, 2004(219) (2004)
Biomarkers and clinical rating scales for sodium pyruvate therapy in patients with mitochondrial disease
Koga Y, et al.
Mitochondrion (2019)
Transcriptome and proteome analysis of soleus muscle of hormone-sensitive lipase-null mice
Hansson O, et al.
Journal of Lipid Research, 46(12), 2614-2623 (2005)
Addition of sodium pyruvate to stored red blood cells attenuates liver injury in a murine transfusion model
Xia S, et al.
Mediators of Inflammation, 2016 (2016)
Embryonic development after follicle culture is influenced by follicle-stimulating hormone isoelectric point range
Vitt UA, et al.
Biology of Reproduction, 65(5) (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service