Skip to Content
Merck
All Photos(3)

Key Documents

N5759

Sigma-Aldrich

4-Nitrophenyl β-D-cellobioside

chromogenic, ≥98% (TLC), powder

Synonym(s):

p-Nitrophenyl β-D-cellobioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO13
CAS Number:
Molecular Weight:
463.39
Beilstein:
100234
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Nitrophenyl β-D-cellobioside, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 49.00-51.00 mg/mL

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@@H]2CO)Oc3ccc(cc3)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12+,13-,14-,15-,16-,17-,18+/m1/s1

InChI key

IAYJZWFYUSNIPN-KFRZSCGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl β-D-cellobioside has been used as a substrate to assay β-D-celluliosidase or cellobiohydrolase (exocellulase) activity in the microbial community. It has also been used as a substrate to study the endoglucanase J30 activity.

Biochem/physiol Actions

4-Nitrophenyl β-D-cellobioside is a cellotriose analog, and a chromogenic substrate commonly used for the detection of cellulase activity. Exoglucanase, endoglucanase and β -glucosidase hydrolyze 4-nitrophenyl β-D-cellobioside to form p-nitrophenol (PNP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas L Ellinghaus et al.
Acta crystallographica. Section D, Structural biology, 74(Pt 7), 702-710 (2018-07-04)
The development of robust enzymes, in particular cellulases, is a key step in the success of biological routes to `second-generation' biofuels. The typical sources of the enzymes used to degrade biomass include mesophilic and thermophilic organisms. The endoglucanase J30 from
Kyung-Min Lee et al.
Journal of microbiology and biotechnology, 21(7), 711-718 (2011-07-28)
A highly efficient cellobiohydrolase (CBH)-secreting basidiomycetous fungus, Agaricus arvensis KMJ623, was isolated and identified based on its morphological features and sequence analysis of internal transcribed spacer rDNA. An extracellular CBH was purified to homogeneity from A. arvencis culture supernatant using
Daniel J Coleman et al.
Analytical biochemistry, 371(2), 146-153 (2007-10-12)
A simple and reliable continuous assay procedure for measurement of cellulase activity from several species using the new substrate resorufin-beta-D-cellobioside (Res-CB) has been developed. The product of enzyme reaction, resorufin, exhibits fluorescence emission at 585 nm with excitation at 571
K Ohmiya et al.
Journal of bacteriology, 161(1), 432-434 (1985-01-01)
An enzyme active against p-nitrophenyl-beta-D-glucoside was purified from logarithmic-phase cells of Ruminococcus albus cultivated in a medium containing ball-milled cellulose. The purification yielded homogeneous enzyme after an approximately 520-fold increase in specific activity and a 9% yield. The enzyme was
G Henriksson et al.
European journal of biochemistry, 259(1-2), 88-95 (1999-01-23)
A 28-kDa endoglucanase was isolated from the culture filtrate of Phanerochaete chrysosporium strain K3 and named EG 28. It degrades carboxymethylated cellulose and amorphous cellulose, and to a lesser degree xylan and mannan but not microcrystalline cellulose (Avicel). EG 28

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service