Skip to Content
Merck
All Photos(5)

Key Documents

L9125

Sigma-Aldrich

L-Leucine-p-nitroanilide

leucine aminopeptidase substrate, chromogenic, ≥98% (TLC), powder

Synonym(s):

L-Leucine-4-nitroanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N3O3
CAS Number:
Molecular Weight:
251.28
Beilstein:
2218433
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

L-Leucine-p-nitroanilide, leucine aminopeptidase substrate

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 50 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](N)C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C12H17N3O3/c1-8(2)7-11(13)12(16)14-9-3-5-10(6-4-9)15(17)18/h3-6,8,11H,7,13H2,1-2H3,(H,14,16)/t11-/m0/s1

InChI key

AXZJHDNQDSVIDR-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Leucine-p-nitroanilide has been used as substrate
  • for the determination of intestinal leucine aminopeptidase activity of juvenile rainbow trout (Oncorhynchus mykiss)
  • in insulin-regulated aminopeptidase inhibition assay
  • for determining exopeptidase activity

Substrates

Substrate for the colorimetric determination of leucine aminopeptidase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidant response and body composition of whiteleg shrimp co-cultured with Nile tilapia in recirculating aquaculture
Sharawy ZZ, et al.
Aquaculture Environment Interactions, 9(3), 257-268 (2017)
Reishi Toshiyama et al.
Oncogene, 38(2), 244-260 (2018-08-10)
Previous studies highlighted that aminopeptidase N (APN)/CD13 acts as a scavenger in the survival of hepatocellular carcinoma (HCC) stem cells by reducing reactive oxygen species (ROS) levels. Hence, it has been proposed that APN/CD13 inhibition can increase cellular ROS levels
Natasa Bozić et al.
Electrophoresis, 26(12), 2476-2480 (2005-05-28)
A general method for detecting leucyl aminopeptidase activity after native polyacrylamide gel electrophoresis (PAGE) in situ is described. The method is based on diazotization of p-nitroaniline, liberated in the polyacrylamide gel by leucyl aminopeptidase action on leucine-p-nitroanilide (LpNA) and subsequent
C Toma et al.
Infection and immunity, 64(11), 4495-4500 (1996-11-01)
The structural gene for the Vibrio cholerae leucine aminopeptidase (lap) was cloned and sequenced. The cloned DNA fragment contained a 1,503-bp open reading frame potentially encoding a 501-amino-acid polypeptide with a calculated molecular mass of 54,442 Da. The deduced amino
Pablo Emiliano Cantón et al.
Frontiers in physiology, 10, 1553-1553 (2020-01-24)
Nezara viridula is a polyphagous stink bug that feeds on crops of economic importance such as corn, soybean and cotton. To increase understanding of the ability of this pest insect to feed on such diverse cropping systems, we analyzed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service