Skip to Content
Merck
All Photos(3)

Key Documents

N3503

Sigma-Aldrich

Nystatin

≥4,400 USP units/mg

Synonym(s):

Fungicidin, Mycostatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51302404
PubChem Substance ID:
NACRES:
NA.85

form

powder

Quality Level

specific activity

≥4,400 USP units/mg

solubility

water: insoluble

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

O[C@@H]([C@H](C)[C@H](C)O1)[C@@H](C)/C=C/C=C/CC/C=C/C=C/C=C/C=C/[C@H](O[C@@H]2O[C@H](C)[C@@H](O)[C@H](N)[C@@H]2O)C[C@@]3([H])[C@H](C(O)=O)[C@@H](O)C[C@](O)(O3)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)C[C@@H](O)CC1=O

InChI

1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28+,29-,30+,31+,32+,33+,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

VQOXZBDYSJBXMA-QEKUPDCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nystatin is a polyene antibiotic.
Chemical structure: polyene

Application

Nystatin is used as a fungal membrane (ergosterol binding) pore forming agent and to create nystatin/ergosterol based ion channels in lipid bilayers. Its uses include preventing cell culture contamination and inducing interleukin (IL)-1, IL-8, and tumor necrosis factor α secretion in TLR2-expressing THP1 cells. This product is recommended for use in cell culture applications at 50 mg/L.

Biochem/physiol Actions

Mode of Action: Nystatin acts by binding to ergosterols in fungal cell membranes, increasing permeability of the membrane, and creating nystatin/ergosterol based ion channels. It also functions as a lipid raft-inhibiting reagent and as membrane associated cholesterol.

Antimicrobial spectrum: Nystatin acts against fungi, yeasts and molds.

Caution

Nystatin in tissue culture media is stable at 37°C for three days. It is also stable in moderately alkaline media. Solutions and aqueous suspensions begin to lose activity soon after preparation, with heat, light, and oxygen accelerating the decomposition. Aqueous suspensions are stable for 10 minutes when heating to 100°C at pH 7.

Preparation Note

Nystatin is soluble at 28°C in methanol (11.2 mg/mL), ethanol (1.2 mg/mL), carbon tetrachloride (1.23 mg/mL), chloroform (0.48 mg/mL), benzene (0.28 mg/mL), and ethylene glycol (8.75 mg/mL). It is also soluble in DMSO at 5 mg/mL and freely soluble in DMF and formamide. It is not recommended to autoclave or sterile filter solutions of Nystatin. Prepare 50 mg/mL stock suspensions in water and store at -20°C.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Store under inert gas. Air and light sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymund R Razonable et al.
Antimicrobial agents and chemotherapy, 49(8), 3546-3549 (2005-07-29)
Nystatin is an antifungal compound with potent proinflammatory properties. Herein, we demonstrate that nystatin induces interleukin (IL)-1beta, IL-8, and tumor necrosis factor alpha secretion through its activation of toll-like receptor 1 (TLR1) and TLR2. Hence, a TLR-dependent mechanism could serve
Santhanam Shanmughapriya et al.
Biofouling, 28(9), 929-935 (2012-09-12)
The presence of intrauterine contraceptive devices (IUDs) gives a solid surface for attachment and an ideal niche for biofilm to form and flourish. Pelvic actinomycosis is often associated with the use of IUDs. Treatment of IUD-associated pelvic actinomycosis requires the
Aikaterini Anagnostopoulou et al.
Scientific reports, 10(1), 17818-17818 (2020-10-22)
Vascular smooth muscle cell (VSMC) function is regulated by Nox-derived reactive oxygen species (ROS) and redox-dependent signaling in discrete cellular compartments. Whether cholesterol-rich microdomains (lipid rafts/caveolae) are involved in these processes is unclear. Here we examined the sub-cellular compartmentalization of
N Akaike et al.
The Japanese journal of physiology, 44(5), 433-473 (1994-01-01)
We have reviewed the basic aspects of history, physicochemical properties, advantages, disadvantages, and applications of nystatin- and amphotericin B-perforated patch recording. Recently, we have developed a novel perforated patch technique using gramicidin [49, 132, 133]. Since gramicidin pores are permeable
Elina Siljamäki et al.
PloS one, 8(2), e55465-e55465 (2013-02-09)
We have previously shown that soluble collagen and a human pathogen, echovirus 1 (EV1) cluster α2β1 integrin on the plasma membrane and cause their internalization into cytoplasmic endosomes. Here we show that cholesterol plays a major role not only in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service