Skip to Content
Merck
All Photos(1)

Key Documents

O8757

Sigma-Aldrich

Ofloxacin

fluoroquinolone antibiotic

Synonym(s):

Dextrofloxacin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
MDL number:
UNSPSC Code:
51282956
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥99% (HPLC)

form

powder

solubility

1 M NaOH: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

FC1=C(C3=[c]4[n](cc([c]([c]4=C1)=O)C(=O)O)C(CO3)C)N2CCN(CC2)C

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)

InChI key

GSDSWSVVBLHKDQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: fluoroquinolone

Application

Ofloxacin is a synthetic fluoroquinolone with broad-spectrum antibacterial activity. It is a nalidixic acid analog. It is given to patients before undergoing retinal reattachment surgery[1].

Biochem/physiol Actions

Ofloxacin inhibits bacterial DNA gyrase and topoisomerase IV, which haults DNA replication and cell division. Ofloxacin has been shown to convert LytA from the inactive E-form to the active C-form[2]. It is a chiral molecule that inhibits pneumococcal cell wall-degrading virulence factors[2].

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Cekiç et al.
The British journal of ophthalmology, 83(10), 1183-1185 (1999-09-30)
To assess the subretinal fluid (SRF) levels of ofloxacin following topical, oral or combined administration. 31 patients undergoing conventional retinal reattachment surgery were randomly assigned to three groups. Nine patients received topical ofloxacin, 11 patients received oral ofloxacin, and the
S Güniz Küçükgüzel et al.
European journal of medicinal chemistry, 42(7), 893-901 (2007-04-10)
Three novel series of 2',4'-difluoro-4-hydroxybiphenyl-3-carboxylic acid derivatives namely 4-substituted-1,2,4-triazoline-3-thiones (4a-g); 2-substituted-1,3,4-thiadiazoles (5a-g) and 2-substituted-1,3,4-oxadiazoles (6a-g) have been synthesized. Twenty-one of the newly synthesized compounds were tested against various bacteria, fungi, yeast species and virus. In addition, we have replaced the
Andrea Von Groll et al.
Antimicrobial agents and chemotherapy, 53(10), 4498-4500 (2009-08-19)
This study evaluated cross-resistance of Mycobacterium tuberculosis strains to ofloxacin, moxifloxacin, and gatifloxacin and investigated the presence of mutations in gyrA and gyrB. Fluoroquinolone susceptibilities were determined for 41 M. tuberculosis strains by the proportion method on 7H11, and MICs
Carlos Fernández-Tornero et al.
The Journal of biological chemistry, 280(20), 19948-19957 (2005-03-17)
The search for new drugs against Streptococcus pneumoniae (pneumococcus) is driven by the 1.5 million deaths it causes annually. Choline-binding proteins attach to the pneumococcal cell wall through domains that recognize choline moieties, and their involvement in pneumococcal virulence makes
O Gutiérrez et al.
Antimicrobial agents and chemotherapy, 51(12), 4329-4335 (2007-10-17)
All (236) Pseudomonas aeruginosa isolates resistant to imipenem and/or meropenem collected during a multicenter (127-hospital) study in Spain were analyzed. Carbapenem-resistant isolates were found to be more frequently resistant to all beta-lactams and non-beta-lactam antibiotics than carbapenem-susceptible isolates (P <

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service