Skip to Content
Merck
All Photos(3)

Key Documents

N1376

Sigma-Aldrich

Naringin

≥90% (HPLC), from citrus fruit

Synonym(s):

4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-neohesperidoside, Naringenine-7-rhamnosidoglucoside, Naringoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
Beilstein:
102012
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

citrus fruit

Assay

≥90% (HPLC)

form

powder

color

white to light yellow

solubility

ethanol: 50 mg/mL, slightly hazy to slightly hazy, light yellow to yellow

storage temp.

room temp

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)CC(Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-7,10,16,18,20-30,32-36H,8-9H2,1H3/t10-,16?,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

DFPMSGMNTNDNHN-JJLSSNRUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naringin is a flavanone glycoside.

Application

Naringin has been used in a study to assess aldosterone production by a human adrenocortical cell line. Long-term naringin consumption reverses a glucose uptake defect and can be used to improve cognitive deficits of Alzheimer′s disease.

Biochem/physiol Actions

Naringin, a flavanoid in grapefruit and other citrus fruits, potently inhibits intestinal organic anion-transporting polypeptide 1A2 (OATP1A2). Grapefruit juice thereby reduces bioavailability of many pharmacological agents taken at the same time.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongmei Wang et al.
Pharmacology, biochemistry, and behavior, 102(1), 13-20 (2012-06-29)
Naringin, a bioflavonoid, has been reported to have potent neuro-protective effects, but its preventive effects on amyloid-β (Aβ) induced, Alzheimer's disease (AD) related, cognitive impairment, and the underlying mechanisms of these effects have not been well characterised. Three-month-old APPswe/PSΔE9 transgenic
Denise C Endringer et al.
Journal of natural products, 71(6), 1082-1084 (2008-05-09)
Aromatase is a well-established target for the chemoprevention of breast cancer. The dihydroisocoumarin (3 R,4 R)-(-)-6-methoxy-1-oxo-3-pentyl-3,4-dihydro-1 H-isochromen-4-yl acetate (1) (IC 50 = 1.6 +/- 0.1 microM), isolated from aerial parts of Xyris pterygoblephara, showed aromatase inhibitory activity. The specificity of
H Serra et al.
Bioorganic & medicinal chemistry, 16(7), 4009-4018 (2008-02-06)
Three glycosilated flavonoids (diosmin, hesperidin and naringin) and respective aglycones were characterized in terms of their apparent ionisation constants and bidirectional permeability using the cellular model Caco-2 as well as the artificial membrane model PAMPA. Ionisation curves were established by
Jiukai Zhang et al.
Food chemistry, 135(3), 1471-1478 (2012-09-08)
Huyou (Citrus changshanensis) is rich in naringin and neohesperidin, which are natural flavanone glycosides with a range of biological activities. Among the different fruit parts, i.e. flavedo, albedo, segment membrane (SM), and juice sacs (JS), albedo showed the highest contents
Yi-Chu Nie et al.
Journal of medicinal food, 15(10), 894-900 (2012-09-19)
Naringin, a well-known flavanone glycoside of grapefruit and citrus fruits, was found to be as an effective anti-inflammatory compound in our previous lipopolysaccharide-induced acute lung injury mouse model via blockading activity of nuclear factor κB. The current study sought to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service